Merck
  • Stearic acids at sn-1, 3 positions of TAG are more efficient at limiting fat deposition than palmitic and oleic acids in C57BL/6 mice.
Stearic acids at sn-1, 3 positions of TAG are more efficient at limiting fat deposition than palmitic and oleic acids in C57BL/6 mice.
The British journal of nutrition (2013-11-30)
Shiou-Wah Gouk, Sit-Foon Cheng, Augustine Soon-Hock Ong, Cheng-Hock Chuah
ABSTRACT
In the present study, we investigated the effect of long-acyl chain SFA, namely palmitic acid (16:0) and stearic acid (18:0), at sn-1, 3 positions of TAG on obesity. Throughout the 15 weeks of the experimental period, C57BL/6 mice were fed diets fortified with cocoa butter, sal stearin (SAL), palm mid fraction (PMF) and high-oleic sunflower oil (HOS). The sn-1, 3 positions were varied by 16:0, 18:0 and 18:1, whilst the sn-2 position was preserved with 18:1. The HOS-enriched diet was found to lead to the highest fat deposition. This was in accordance with our previous postulation. Upon normalisation of total fat deposited with food intake to obtain the fat:feed ratio, interestingly, mice fed the SAL-enriched diet exhibited significantly lower visceral fat/feed and total fat/feed compared with those fed the PMF-enriched diet, despite their similarity in SFA-unsaturated fatty acid-SFA profile. That long-chain SFA at sn-1, 3 positions concomitantly with an unsaturated FA at the sn-2 position exert an obesity-reducing effect was further validated. The present study is the first of its kind to demonstrate that SFA of different chain lengths at sn-1, 3 positions exert profound effects on fat accretion.
Product Description
Supelco
Sunflower seed oil, analytical standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Stearic acid, analytical standard
Supelco
Oleic acid, analytical standard
Supelco
Palm oil, analytical standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sunflower seed oil from Helianthus annuus
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Supelco
Oleic acid, Selectophore, ≥99%
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Palmitic acid, analytical standard
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Glyceryl trioleate, ≥97.0% (TLC)
Sigma-Aldrich
Glyceryl trioleate, ≥99%
Sigma-Aldrich
Glyceryl trioleate, ~65%
Sigma-Aldrich
Palmitic acid, ≥99%
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard