Skip to Content
Merck
All Photos(1)

Key Documents

T7140

Sigma-Aldrich

Glyceryl trioleate

≥99%

Synonym(s):

(9Z)9-Octadecenoic acid 1,2,3-propanetriyl ester, 1,2,3-Tri(cis-9-octadecenoyl)glycerol, Glycerol trioleate, Glycerol triolein, Oleic acid triglyceride, Oleic triglyceride, TG(18:1(9Z)/18:1(9Z)/18:1(9Z)), Triolein

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H33COOCH2)2CHOCOC17H33
CAS Number:
Molecular Weight:
885.43
Beilstein:
1718692
EC Number:
MDL number:
UNSPSC Code:
41141833
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (sunflower)

Assay

≥99%

form

oil

bp

235-240 °C/18 mmHg (lit.)

density

0.91 g/mL (lit.)

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]C(COC(CCCCCCC/C=C\CCCCCCCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-

InChI key

PHYFQTYBJUILEZ-IUPFWZBJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glyceryl trioleate is derived from glycerol. It is composed of three oleic acid units and is an unsaturated triglyceride.

Application

Glyceryl trioleate has been used:
  • as an experimental diet along with fat-free basal mix and corn oil and then to access the dietary fat absorption among mice
  • as an interfering substance to test its effect on human serum in the approach to develop rapid enzyme immunoassay for the detection of retinol-binding protein
  • as a standard in the determination of triglyceride concentration, colorimetrically using liver tissue sample from cows

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

626.0 °F - closed cup

Flash Point(C)

330.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stearoyl-CoA desaturase-1 mediates the pro-lipogenic effects of dietary saturated fat
Sampath H, et al.
The Journal of Biological Chemistry, 282(4), 2483-2493 (2007)
Sawsan Amara et al.
Scientific reports, 6, 24985-24985 (2016-04-30)
Oleaginous microorganisms have considerable potential for biofuel and commodity chemical production. Under nitrogen-limitation, Rhodococcus jostii RHA1 grown on benzoate, an analog of lignin depolymerization products, accumulated triacylglycerols (TAGs) to 55% of its dry weight during transition to stationary phase, with
Chemistry and Physics for Nurse Anesthesia, 327-327 (2009)
Effect of glycerol and its analogs on polyhydroxyalkanoate biosynthesis by recombinant Ralstonia eutropha: a quantitative structure-activity relationship study of chain transfer agents
Tsuge T, et al.
Polymer Degradation and Stability, 98(9), 1586-1590 (2013)
Energy balance, metabolic status, and the first postpartum ovarian follicle wave in cows administered propylene glycol
Butler ST, et al.
Journal of Dairy Science, 89(8), 2938-2951 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service