Skip to Content
Merck
All Photos(1)

Documents

76119

Supelco

Palmitic acid

analytical standard

Synonym(s):

1-Pentadecanecarboxylic acid, C16:0, Cetylic acid, Hexadecanoic acid, NSC 5030, PamOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOH
CAS Number:
Molecular Weight:
256.42
Beilstein:
607489
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

10 mmHg ( 210 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

271.5 °C/100 mmHg (lit.)

mp

61-62.5 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)

InChI key

IPCSVZSSVZVIGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Palmitic acid belongs to the group of saturated fatty acids and accounts for 20-30 % of the total fatty acids present in the human body.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

This analytical standard can also be used as follows:
  • Development of a non-destructive Vis-NIR reflectance spectroscopy-based method for the detection of four fatty acids in 93 samples of seven different edibles oils
  • Simultaneous identification and quantification of fatty acids from blood plasma samples after their methyl derivatization by large-volume injection (LVI) gas chromatography combined with vacuum ultraviolet spectroscopy (GC-VUV)
  • Determination of the fatty acid composition of sunflower seed oils extracted from 93 different sunflower varieties by near-infrared reflectance spectroscopy (NIRS) combined with modified partial least squares (MPLS) and partial least squares (PLS) regression methods
  • Separation and quantification of palmitic, oleic, linoleic, and linolenic acids from 186 samples of sea buckthorn seed oil using near-infrared spectroscopy (NIR) in combination with four different combinations of multivariate calibration methods— partial least squares (PLS), PLS-uninformative variables elimination (UVE), PLS- competitive adaptive reweighted sampling (CARS), and multiple linear regression (MLR)-UVE
  • Quantitative analysis of various bioactive compounds from the ethanolic extract of sesame seeds by gas chromatography-mass spectrometry (GC-MS)
  • Multi-residue analysis of oleic acid and other related fatty acids using a derivatization-free method based on gas chromatography-flame ionization detection (GC-FID)
  • Ultra-performance liquid chromatography-mass spectrometry (UHPLC) method-based separation, identification, and determination of eight saturated and unsaturated fatty acids in eight different commercial samples of olive oil without any prior sample treatment
  • Development of a liquid chromatography-mass spectrometry (LC-MS) method to measure seven fatty acid esters of hydroxy fatty acids (FAHFAs), four fatty acids, and four cardiovascular-related biomarkers in plasma samples of 57 healthy individuals

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of myristic acid versus palmitic acid on serum lipid and lipoprotein levels in healthy women and men
Zock LP, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 14(4), 567-575 (1994)
Detection of unusual very-long-chain fatty acid and ether lipid derivatives in the fibroblasts and plasma of patients with peroxisomal diseases using liquid chromatography-mass spectrometry
Takashima S, et al.
Molecular Genetics and Metabolism, 120(3), 255-268 (2017)
B Vessby et al.
Diabetologia, 37(10), 1044-1050 (1994-10-01)
Recent data indicate that peripheral insulin sensitivity may be influenced by dietary fat quality and skeletal muscle phospholipid fatty acid composition. During a health survey of 70-year-old men insulin sensitivity was measured by the euglycaemic hyperinsulinaemic clamp technique and the
Jonas Sieber et al.
The American journal of pathology, 183(3), 735-744 (2013-07-23)
Type 2 diabetes mellitus is characterized by dyslipidemia with elevated free fatty acids (FFAs). Loss of podocytes is a hallmark of diabetic nephropathy, and podocytes are highly susceptible to saturated FFAs but not to protective, monounsaturated FFAs. We report that
Li Liu et al.
Experimental neurology, 248, 482-490 (2013-08-24)
Deregulation of calcium has been implicated in neurodegenerative diseases, including Alzheimer's disease (AD). Previously, we showed that saturated free-fatty acid, palmitate, causes AD-like changes in primary cortical neurons mediated by astrocytes. However, the molecular mechanisms by which conditioned medium from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service