Skip to Content
Merck
All Photos(1)

Key Documents

47123

Supelco

Sunflower seed oil

analytical standard

Synonym(s):

Sunflower seed oil from Helianthus annuus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Characterized sunflower seed oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses. These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen. A Certificate of Composition, which includes the chromatographic fingerprint analysis, is provided with the oil sample.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Prieto et al.
Journal of animal science, 91(1), 446-454 (2012-10-26)
The aim of this study was to investigate the effects of milking frequency reduction and dietary lipid supplementation on intake, BW, and milk yield and composition in high yielding dairy ewes. Ten lactating Assaf ewes were allocated into 2 experimental
M A De la Rubia et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(2), 410-417 (2012-11-22)
Due to the chemical and physical structure of a lignocellulosic biomass, its anaerobic digestion (AD) is a slow and difficult process. In this paper, the results obtained from a batch biochemical methane potential (BMP) test and fed-batch mesophilic AD assays
Liliana Albertengo et al.
Carbohydrate polymers, 92(2), 1641-1646 (2013-02-13)
Chitosan is a modified, natural carbohydrate polymer derived by deacetylation of chitin. Due to the presence of two functional groups can undergo many chemical modifications. In a previous work we described the synthetic strategy and characterization of a novel soluble
Yun-Sang Choi et al.
Meat science, 93(3), 652-658 (2013-01-01)
The effects of reducing pork fat levels from 30% to 20% by partially substituting pork fat with a mix of sunflower seed oil (0, 5, 10, 15, and 20%) and makgeolli lees fiber (2%) were investigated based on physicochemical properties
Carlos Ferreiro-Vera et al.
Food chemistry, 136(2), 576-584 (2012-11-06)
The effect of breakfast intake of fried oils containing natural antioxidants or a synthetic autooxidation inhibitor on the metabolism of essential fatty acids focused on obese individuals. Serum levels of eicosanoids were compared in individuals before and after intake of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service