Skip to Content
Merck
All Photos(1)

Key Documents

O1008

Sigma-Aldrich

Oleic acid

≥99% (GC)

Synonym(s):

9Z-Octadecenoic acid, cis-Oleic acid, cis-9-Octadecenoic acid, Elainic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein:
1726542
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Sunflower)

Quality Level

vapor pressure

1 mmHg ( 176 °C)

Assay

≥99% (GC)

form

viscous liquid

color

colorless to very faintly yellow

refractive index

n20/D 1.459 (lit.)

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

solubility

H2O: insoluble (practically)
alcohol: soluble
benzene: soluble
chloroform: soluble
oil: soluble (fixed and volatile)

density

0.89 g/mL at 25 °C (lit.)

application(s)

life science and biopharma
lipidomics

functional group

oleic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

ZQPPMHVWECSIRJ-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Oleic acid (OA), a non-essential fatty acid, is classified as monounsaturated fatty acid (MUFA) richly present in olive oil. Its quantitative removal from the mixtures of triglyceride and partial glycerides has been reported. OA is widely used as a surfactant for the stabilization of magnetic nanoparticles. Monodisperse magnetite nanoparticles (7 and 19nm) coated with OA have been prepared.

Application

Oleic acid has been used:
  • to study the effects of fatty acids on gastric cancer (GC)
  • to induce lipid droplet formation in cell culture
  • as an additive in the magnetorheological fluid preparation.
Oleic acid may be used in the preparation of 10-hydroxy octadecanoic acid and methyl ester of the 10-hydroxy octadecanoic acid.

Biochem/physiol Actions

Oleic acid is known to improve immune function and health. Dietary supplement of this fatty acid contributes to the reduction in cholesterol levels, atherogenesis risk, blood pressure, and host versus graft response. Oleic acid has been proven to benefit autoimmune diseases by exhibiting anti-inflammatory action. It also ameliorates the condition of breast cancer. Oleic acid produces physiological effects such as protein kinase C activation in hepatocytes, uncoupling oxidative phosphorylation, and 2,4-dinitrophenol−stimulated ATPase inhibition. This action can be reversed by adding serum albumin.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE STEREOSPECIFIC CONVERSION OF STEARIC ACID TO OLEIC ACID.
G J SCHROEPFER et al.
The Journal of biological chemistry, 240, 54-63 (1965-01-01)
CD36 mediates palmitate acid-induced metastasis of gastric cancer via AKT/GSK-3?/?-catenin pathway
Pan J, et al.
Journal of Experimental & Clinical Cancer Research, 38 (2019)
Oleic acid coating on the monodisperse magnetite nanoparticles.
Zhang L, et al.
Applied Surface Science, 253(5), 2611-2617 (2006)
Influence of graphite flakes on the strength of magnetorheological fluids at high temperature and its rheology
Thakur MK and Sarkar C
IEEE Transactions on Magnetics, 56(5) (2020)
Kun Huang et al.
Journal of hepatology, 66(5), 962-977 (2016-12-17)
PARP1 is a key mediator of cellular stress responses and critical in multiple physiological and pathophysiological processes of cells. However, whether it is involved in the pathogenesis of non-alcoholic fatty liver disease (NAFLD) remains elusive. We analysed PARP1 activity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service