Skip to Content
Merck
All Photos(1)

Documents

S4751

Sigma-Aldrich

Stearic acid

Grade I, ≥98.5% (capillary GC)

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein:
608585
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

vapor pressure

1 mmHg ( 173.7 °C)

type

Grade I

Assay

≥98.5% (capillary GC)

form

powder

bp

361 °C (lit.)

mp

67-72 °C (lit.)

solubility

chloroform: 100 mg/mL, clear, colorless

density

0.845 g/cm3

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Stearic acid is a C18 saturated fatty acid.

Application

Stearic acid has been used in fatty acid preparations.

Biochem/physiol Actions

β-Oxidation of stearic acid yields eight FADH2 (flavin adenine dinucleotide) and NADH2 (nicotinamide adenine dinucleotide) molecules and nine acetyl-CoA molecules.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

392.0 °F - Cleveland open cup

Flash Point(C)

200 °C - Cleveland open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxia potentiates palmitate-induced pro-inflammatory activation of primary human macrophages
Snodgrass Ryan G, et al.
The Journal of Biological Chemistry, 291(1), 413-424 (2016)
Construction and comparison of solid liquid phase diagrams of binary sat-sat and unsat-unsat fatty acid mixtures comprising two of palmitic/stearic/oleic/linoleic/linolenic acid
Eckert KA, et al.
Journal of Environmental Research And Development, 10(4), 1-1 (2016)
Solid liquid phase diagrams of binary fatty acid mixtures?Palmitic/stearic with oleic/linoleic/linolenic acid mixture
Eckert Karl-Andreas, et al.
Thermochimica Acta, 630, 50-63 (2016)
Sports Nutrition (1999)
Hua Ning et al.
Nutrients, 8(4), 227-227 (2016-04-23)
The detrimental role of hepatic lipotoxicity has been well-implicated in the pathogenesis of NAFLD. Previously, we reported that inhibiting autophagy aggravated saturated fatty acid (SFA)-induced hepatotoxicity. Insulin, a physiological inhibitor of autophagy, is commonly increased within NAFLD mainly caused by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service