Skip to Content
Merck
All Photos(1)

Key Documents

T7752

Sigma-Aldrich

Glyceryl trioleate

~65%

Synonym(s):

(9Z)9-Octadecenoic acid 1,2,3-propanetriyl ester, 1,2,3-Tri(cis-9-octadecenoyl)glycerol, Glycerol trioleate, Glycerol triolein, Oleic acid triglyceride, Oleic triglyceride, TG(18:1(9Z)/18:1(9Z)/18:1(9Z)), Triolein

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H33COOCH2)2CHOCOC17H33
CAS Number:
Molecular Weight:
885.43
Beilstein:
1718692
EC Number:
MDL number:
UNSPSC Code:
41141833
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

form

liquid

concentration

~65%

bp

235-240 °C/18 mmHg (lit.)

density

0.91 g/mL (lit.)

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

2-8°C

SMILES string

[H]C(COC(CCCCCCC/C=C\CCCCCCCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-

InChI key

PHYFQTYBJUILEZ-IUPFWZBJSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

626.0 °F - closed cup

Flash Point(C)

330.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina M E Bitar et al.
Biotechnology progress, 35(6), e2851-e2851 (2019-05-28)
In this study, we developed a high-throughput microchannel emulsification process to encapsulate pancreatic beta cells in monodisperse alginate beads. The process builds on a stirred emulsification and internal gelation method previously adapted to pancreatic cell encapsulation. Alginate bead production was
Jianfeng Tang et al.
Environmental pollution (Barking, Essex : 1987), 164, 158-163 (2012-03-01)
Triolein embedded cellulose acetate membrane (TECAM) passive samplers provide potentially powerful tool for monitoring time weighted average concentrations (C(TWA)) of trace hydrophobic organic contaminants in water. To study the field performance of TECAM, exchange kinetics of chemicals between water and
Mira Rosenblat et al.
BioFactors (Oxford, England), 38(4), 292-299 (2012-06-08)
We analyzed, for the first time, both in vitro and in vivo, the effect of very low density lipoprotein (VLDL), or of pure triglycerides, on high-density lipoprotein (HDL)-associated paraoxonase1 (PON1) catalytic activities. Incubation of serum or HDL from healthy subjects
Mi-Hwa Lee et al.
Applied and environmental microbiology, 78(14), 4959-4966 (2012-05-01)
A novel lipolytic enzyme was isolated from a metagenomic library obtained from tidal flat sediments on the Korean west coast. Its putative functional domain, designated MPlaG, showed the highest similarity to phospholipase A from Grimontia hollisae CIP 101886, though it
Sylvie Hollebeeck et al.
Food chemistry, 138(2-3), 1936-1944 (2013-02-16)
Bioaccessibility studies should be taken into account when evaluating the physiological effects of ingested compounds at the intestine level. Several in vitro digestion protocols have been described, with a wide range of experimental conditions but no optimised protocol exists. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service