Skip to Content
Merck
Sulfuric acid puriss., meets analytical specification of Ph. Eur., BP, 95-97%

Sulfuric acid

Sulfuric acid puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Synonym(s):
Sulfuric acid, Sulfuric acid solution
Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Compatibility
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-20 of 22
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2