If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
1.00731
Sulfuric acid
95-97%, for analysis EMSURE® ISO
Synonym(s):
Sulfuric acid
About This Item
Recommended Products
Product Name
Sulfuric acid 95-97%, for analysis EMSURE® ISO
Agency
reag. ISO
Quality Level
vapor density
<0.3 (25 °C, vs air)
vapor pressure
0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)
product line
EMSURE®
Assay
95-97% (alkalimetric)
form
liquid
pH
0.3 (25 °C, 49 g/L in H2O)
bp
~290 °C (lit.)
mp
-20 °C
density
1.840 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤0.1 ppm
nitrate (NO3-): ≤0.2 ppm
phosphate (PO43-): ≤0.5 ppm
cation traces
Ag: ≤0.010 ppm
Al: ≤0.050 ppm
As: ≤0.010 ppm
Au: ≤0.050 ppm
B: ≤0.050 ppm
Ba: ≤0.050 ppm
Be: ≤0.010 ppm
Bi: ≤0.050 ppm
Ca: ≤0.1 ppm
Cd: ≤0.01 ppm
Co: ≤0.01 ppm
Cr: ≤0.020 ppm
Cu: ≤0.01 ppm
Fe: ≤0.1 ppm
Ga: ≤0.05 ppm
Ge: ≤0.02 ppm
In: ≤0.05 ppm
K: ≤0.1 ppm
Li: ≤0.01 ppm
Mg: ≤0.05 ppm
Mn: ≤0.01 ppm
Mo: ≤0.02 ppm
NH4+: ≤1 ppm
Na: ≤0.3 ppm
Ni: ≤0.02 ppm
Pb: ≤0.01 ppm
Pt: ≤0.1 ppm
Sn: ≤0.05 ppm
Sr: ≤0.01 ppm
Ti: ≤0.02 ppm
Tl: ≤0.02 ppm
V: ≤0.01 ppm
Zn: ≤0.05 ppm
Zr: ≤0.02 ppm
SMILES string
OS(O)(=O)=O
storage temp.
no temp limit
InChI
1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)
InChI key
QAOWNCQODCNURD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- A systematic investigation of key factors of nucleic acid precipitation toward optimized DNA/RNA isolation.: This research examines the use of 2-Propanol in the precipitation of nucleic acids, optimizing conditions for DNA and RNA isolation procedures in molecular biology (Li et al., 2020).
- Staining of RNA and DNA on electrophoretic gels and in cytology with juice of Vaccinium myrtillus berries.: Utilizes 2-Propanol in the staining process of nucleic acids on gels, enhancing visibility and differentiation of RNA and DNA bands (Ahokas, 2019).
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
What is the difference between 1007311011 and 1007311000
1 answer-
Product 1007311011 is packaged in a HPDE bottle while product 1007311000 is packaged in a amber glass bottle.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service