Skip to Content
Merck
All Photos(1)

Key Documents

07208

Sigma-Aldrich

Sulfuric acid

puriss., meets analytical specification of Ph. Eur., BP, 95-97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

<0.3 (25 °C, vs air)

Quality Level

vapor pressure

1 mmHg ( 146 °C)

description

Nominally 95-98% H2SO4

grade

puriss.

Assay

95-97%

form

liquid

quality

meets analytical specification of Ph. Eur., BP

impurities

≤0.0002% KMnO4 red. matter (as O)
≤0.0005% heavy metals (as Pb)

ign. residue

≤0.0005%

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
nitrate (NO3-): ≤1 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

complies for appearance of solution

SMILES string

OS(O)(=O)=O
OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Scanning electrochemical microscopy for the differentiation of radical-induced degradation mechanisms in polymer electrolyte membranes.: This study employs sulfuric acid in scanning electrochemical microscopy to differentiate radical-induced degradation mechanisms in polymer electrolyte membranes, crucial for fuel cell development (Thiel & Eichelbaum, 2024).

  • Formation of Chromophores from cis-Pinonaldehyde Aged in Highly Acidic Conditions.: Investigates the formation of chromophores from cis-Pinonaldehyde under highly acidic conditions, contributing to our understanding of atmospheric chemistry and the role of sulfuric acid (Wong et al., 2024).

  • Optimizing cellulose fraction for enhanced utility: Comparative pre-treatment of Agave tequilana Weber var. blue bagasse fiber for sustainable applications.: This research focuses on optimizing the cellulose fraction using sulfuric acid pre-treatment to enhance the utility of Agave tequilana fibers for sustainable applications (Lazaro-Romero et al., 2024).

  • Studying the Structure and Properties of Epoxy Composites Modified by Original and Functionalized with Hexamethylenediamine by Electrochemically Synthesized Graphene Oxide.: Explores the use of sulfuric acid in the synthesis of graphene oxide for modifying epoxy composites, enhancing their structural and mechanical properties (Mostovoy et al., 2024).

  • Inhibition of corrosion on API 5L X52 pipeline steel in acid media by Tradescantia spathacea.: This study examines the effectiveness of Tradescantia spathacea as a corrosion inhibitor for steel in sulfuric acid media, significant for industrial applications (Rodríguez-Torres et al., 2024).

Packaging

In PE-bottles; also available in Glass-bottles (Material no. ending with *-GL), PE canister (Material no. ending with *-H)

Other Notes

The article number 07208-4X2.5L-M will be discontinued. Please order the single bottle 07208-2.5L-M which is physically identical with the same exact specifications.
The article number 07208-6X1L will be discontinued. Please order the single bottle 07208-1L which is physically identical with the same exact specifications.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Li et al.
Journal of chromatography. A, 1404, 39-43 (2015-06-09)
The paper reports on a novel method for rapid determination of carbonyl in lignins by headspace gas chromatography (HS-GC). The method involves the quantitative carbonyl reduction for aldehydes in 2min at room temperature or for acetones in 30min at 80°C
Investigation of the kinetics and mechanism of the anodic oxidation of aniline in aqueous sulfuric acid solution at a platinum electrode.
Mohilner DM, et al.
Journal of the American Chemical Society, 84(19), 3618-3622 (1962)
Self-Ordering of Cell Arrangement of Anodic Porous Alumina Formed in Sulfuric Acid Solution.
Masuda H, et al.
Journal of the Electrochemical Society, 144(5), L127-L130 (1997)
Francesco Riccobono et al.
Science (New York, N.Y.), 344(6185), 717-721 (2014-05-17)
Atmospheric new-particle formation affects climate and is one of the least understood atmospheric aerosol processes. The complexity and variability of the atmosphere has hindered elucidation of the fundamental mechanism of new-particle formation from gaseous precursors. We show, in experiments performed
João Almeida et al.
Nature, 502(7471), 359-363 (2013-10-08)
Nucleation of aerosol particles from trace atmospheric vapours is thought to provide up to half of global cloud condensation nuclei. Aerosols can cause a net cooling of climate by scattering sunlight and by leading to smaller but more numerous cloud

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service