Skip to Content
Merck
All Photos(2)

Key Documents

1.00714

Supelco

Sulfuric acid

96%, Suprapur®

Synonym(s):

Sulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-639-5
NACRES:
NA.21

Product Name

Sulfuric acid 96%, for inorganic trace analysis, Suprapur®

grade

for inorganic trace analysis

Quality Level

Agency

suitable for EPA 300

vapor density

<0.3 (25 °C, vs air)

vapor pressure

0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)

Assay

≥95% (acidimetric)
96%

form

liquid

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤100 ppb
nitrate (NO3-): ≤200 ppb
phosphate (PO43-): ≤100 ppb

cation traces

Ag: ≤1.0 ppb
Al: ≤5.0 ppb
As: ≤0.5 ppb
Au: ≤0.5 ppb
Ba: ≤1.0 ppb
Be: ≤0.5 ppb
Bi: ≤0.5 ppb
Ca: ≤5.0 ppb
Cd: ≤1.0 ppb
Co: ≤0.5 ppb
Cr: ≤1.0 ppb
Cu: ≤0.5 ppb
Fe: ≤5.0 ppb
Ga: ≤0.5 ppb
Ge: ≤1.0 ppb
Hg: ≤2.0 ppb
In: ≤0.5 ppb
K: ≤2.0 ppb
Li: ≤0.5 ppb
Mg: ≤5.0 ppb
Mn: ≤0.5 ppb
Mo: ≤0.5 ppb
NH4+: ≤1.0 ppm
Na: ≤10.0 ppb
Ni: ≤2.0 ppb
Pb: ≤1.0 ppb
Pt: ≤0.5 ppb
Sb: ≤1.0 ppb
Se: ≤1.0 ppb
Sn: ≤1.0 ppb
Sr: ≤0.5 ppb
Ti: ≤1.0 ppb
Tl: ≤0.5 ppb
V: ≤0.5 ppb
Zn: ≤2.0 ppb
Zr: ≤0.5 ppb

SMILES string

OS(O)(=O)=O

storage temp.

no temp limit

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric): ≥ 95.0 %
Colour: ≤ 10 Hazen
Chloride (Cl): ≤ 100 ppb
Nitrate (NO₃): ≤ 200 ppb
Phosphate (PO₄): ≤ 100 ppb
Ag (Silver): ≤ 1.0 ppb
Al (Aluminium): ≤ 5.0 ppb
As (Arsenic): ≤ 0.5 ppb
Au (Gold): ≤ 0.5 ppb
Ba (Barium): ≤ 1.0 ppb
Be (Beryllium): ≤ 0.5 ppb
Bi (Bismuth): ≤ 0.5 ppb
Ca (Calcium): ≤ 5.0 ppb
Cd (Cadmium): ≤ 1.0 ppb
Co (Cobalt): ≤ 0.5 ppb
Cr (Chromium): ≤ 1.0 ppb
Cu (Copper): ≤ 0.5 ppb
Fe (Iron): ≤ 5.0 ppb
Ga (Gallium): ≤ 0.5 ppb
Ge (Germanium): ≤ 1.0 ppb
Hg (Mercury): ≤ 2.0 ppb
In (Indium): ≤ 0.5 ppb
K (Potassium): ≤ 2.0 ppb
Li (Lithium): ≤ 0.5 ppb
Mg (Magnesium): ≤ 5.0 ppb
Mn (Manganese): ≤ 0.5 ppb
Mo (Molybdenum): ≤ 0.5 ppb
NH₄ (Ammonium): ≤ 1.0 ppm
Na (Sodium): ≤ 10.0 ppb
Ni (Nickel): ≤ 2.0 ppb
Pb (Lead): ≤ 1.0 ppb
Pt (Platinum): ≤ 0.5 ppb
Sb (Antimony): ≤ 1.0 ppb
Se (Selenium): ≤ 1.0 ppb
Sn (Tin): ≤ 1.0 ppb
Sr (Strontium): ≤ 0.5 ppb
Ti (Titanium): ≤ 1.0 ppb
Tl (Thallium): ≤ 0.5 ppb
V (Vanadium): ≤ 0.5 ppb
Zn (Zinc): ≤ 2.0 ppb
Zr (Zirconium): ≤ 0.5 ppb
Substances reducing potassium permanganate (as SO₂): ≤ 2 ppm
Residue on ignition: ≤ 2 ppm
The actual values are subject to unavoidable systematic
variations in this concentration range.

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service