Skip to Content
Merck

1.00748

Sulfuric acid

98%, for the determination of nitrogen

Synonym(s):

Sulfuric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
UNSPSC Code:
12352301
EC Index Number:
231-639-5
NACRES:
NA.21
Beilstein/REAXYS Number:
2037554
MDL number:
Assay:
≥97.5% (alkalimetric)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sulfuric acid 98%, for the determination of nitrogen

vapor density

<0.3 (25 °C, vs air)

Quality Segment

vapor pressure

0.0001 hPa ( 20 °C), 1 mmHg ( 146 °C)

assay

≥97.5% (alkalimetric)

form

liquid

ign. residue

≤2 ppm

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

solubility

water: soluble at 20 °C

density

1.840 g/mL at 25 °C (lit.)

cation traces

N: ≤1 ppm

storage temp.

no temp limit

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Application

Sulfuric acid 98% can be used in the Kjeldahl method for the determination of nitrogen.

Analysis Note

Assay (alkalimetric): ≥ 97.5 %
Total nitrogen (N): ≤ 1 ppm
Residue on ignition: ≤ 2 ppm
Date of expiry: see product label


Still not finding the right product?

Explore all of our products under Sulfuric acid 98%


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
100748250004022536010045
100748050004022536010021
100748902504022536555515