Skip to Content
Merck
All Photos(2)

Key Documents

258105

Sigma-Aldrich

Sulfuric acid

ACS reagent, 95.0-98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

suitable for EPA 1613
suitable for SM 4500 - NH3
suitable for SM 5210

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

description

Free from suspended or insoluble matter
Nominally 95-98% H2SO4

Assay

95.0-98.0%

form

liquid

ign. residue

≤5 ppm

color

APHA: ≤10

pH

1.2 (5 g/L)

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

anion traces

MnO4- reducers: ≤2 ppm
chloride (Cl-): ≤0.2 ppm
nitrate (NO3-): ≤0.5 ppm

cation traces

As: ≤0.01 ppm
Fe: ≤0.2 ppm
Hg: ≤5 ppb
NH4+: ≤2 ppm
heavy metals (as Pb): ≤1 ppm

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfuric acid can be used as a catalyst:
  • In the hydrolysis of primary or secondary nitroalkane salts to aldehydes or ketones.
  • In the selective hydrolysis of nitriles to amides.
  • To synthesize amides of t-alkylcarbinamines via Ritter reaction between nitriles and alkenes.
  • To prepare 3,4-dialkyl-2(5H)-furanones by carbonylation of α,β-unsaturated aldehydes.
  • In the nitration of aromatic carbocycles and heteroaromatic compounds.
  • In the Friedel–Craft ketone synthesis using anhydrides via α-amidoalkylation reaction.

H2SO4 can also be used in the following reactions:
  • Michael′s addition reactions
  • Electrophilic substitution reactions
  • Esterification of highly hindered aromatic acids
  • Aldol condensation reaction
  • Beckmann rearrangement
  • Hofmann–Loffler–Freytag reaction
  • Isomerization
  • Dehydrogenation
  • Sulfonation

Other Notes

ACS specifications same as product 320501
4L and 4X4L are packaged in poly bottles
SKU-Pack sizes with -PC are packaged in PVC coated glass bottles.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tim Olding et al.
Physics in medicine and biology, 56(5), 1259-1279 (2011-02-03)
This work develops imaging protocols for improved dose readout of a Fricke-xylenol orange-gelatin (FXG) gel-filled 1 L polyethylene terephthalate (PETE) jar dosimeter using a commercial Vista(TM) cone-beam optical computed tomography (CT) scanner from Modus Medical Devices Inc. (London, ON, Canada).
Determination of mercury in natural waters and effluents by flameless atomic absorption spectrophotometry.
S H Omang
Analytica chimica acta, 53(2), 415-419 (1971-02-01)
Jing Li et al.
Journal of chromatography. A, 1404, 39-43 (2015-06-09)
The paper reports on a novel method for rapid determination of carbonyl in lignins by headspace gas chromatography (HS-GC). The method involves the quantitative carbonyl reduction for aldehydes in 2min at room temperature or for acetones in 30min at 80°C
Investigation of the kinetics and mechanism of the anodic oxidation of aniline in aqueous sulfuric acid solution at a platinum electrode.
Mohilner DM, et al.
Journal of the American Chemical Society, 84(19), 3618-3622 (1962)
Steven Babic et al.
Physics in medicine and biology, 53(6), 1637-1650 (2008-03-28)
Freshly prepared radiochromic ferrous xylenol-orange (FX) gels optically scanned with a light source exhibit a threshold dose response that is thermally and wavelength dependent. Correction for this threshold dose leads to accurate dose calibration and better reproducibility in multiple fraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service