Skip to Content
Merck
All Photos(2)

Key Documents

1.01833

Supelco

Sulfuric acid

95-97%, for analysis EMPARTA® ACS

Synonym(s):

Sulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-639-5
NACRES:
NA.21

Product Name

Sulfuric acid 95-97%, for analysis EMPARTA® ACS

grade

ACS reagent

Quality Level

vapor density

<0.3 (25 °C, vs air)

vapor pressure

0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)

product line

EMPARTA®

Assay

95-97% (alkalimetric)

form

liquid

impurities

≤2 ppm Substances reducing potassium permanganate (as SO2)

ign. residue

≤5 ppm

color

APHA: ≤10

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.2 ppm
nitrate (NO3-): ≤0.5 ppm

cation traces

As: ≤0.01 ppm
Fe: ≤0.2 ppm
Hg: ≤0.005 ppm
NH4+: ≤2 ppm
heavy metals (as Pb): ≤1 ppm

storage temp.

no temp limit

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (alkalimetric): 95.0 - 97.0 %
Identity: passes test
Appearance: passes test
Color: ≤ 10 Hazen
Chloride (Cl): ≤ 0.2 ppm
Nitrate (NO₃): ≤ 0.5 ppm
Heavy metals (as Pb): ≤ 1 ppm
As (Arsenic): ≤ 0.01 ppm
Fe (Iron): ≤ 0.2 ppm
Hg (Mercury): ≤ 0.005 ppm
NH₄ (Ammonium): ≤ 2 ppm
Substances reducing potassium permanganate (as SO₂): ≤ 2 ppm
Residue on ignition: ≤ 5 ppm

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service