Skip to Content
Merck
All Photos(1)

Key Documents

30743-M

Sigma-Aldrich

Sulfuric acid

Sulfuric acid
1 of 1 reviewers received a sample product or took part in a promotion

puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

description

Nominally 95-98% H2SO4

Assay

95.0-97.0%

form

liquid

impurities

≤1 ppm heavy metals (as Pb)
≤2 ppm KMnO4 red. matter (as O)
≤2 ppm ammonium (NH4)

ign. residue

≤5 ppm

color

APHA: ≤10

pH

1.2 (5 g/L)

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.1 ppm
nitrate (NO3-): ≤0.2 ppm
phosphate (PO43-): ≤0.00005%

cation traces

Ag: ≤0.000002%
Al: ≤0.000005%
As: ≤0.01 ppm
Ba: ≤0.000005%
Be: ≤0.000001%
Bi: ≤0.000005%
Ca: ≤0.00002%
Cd: ≤0.000002%
Co: ≤0.000001%
Cr: ≤0.000005%
Cu: ≤0.000001%
Fe: ≤0.1 ppm
Ge: ≤0.000005%
Hg: ≤0.005 ppm
K: ≤0.00001%
Li: ≤0.000001%
Mg: ≤0.000005%
Mn: ≤0.000001%
Mo: ≤0.000002%
Na: ≤0.00005%
Ni: ≤0.000002%
Pb: ≤0.000002%
Sr: ≤0.000002%
Ti: ≤0.00001%
Tl: ≤0.000005%
V: ≤0.000001%
Zn: ≤0.000005%
Zr: ≤0.1 mg/kg

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfuric acid can be used in the colorimetric determination of mercury in water samples.

Packaging

In PE-bottles;also available PE-bottles black (Material no. ending with PEBL; Glass-bottles (Material no. ending with *-GL), PE canister (Material no. ending with *-H)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Li et al.
Journal of chromatography. A, 1404, 39-43 (2015-06-09)
The paper reports on a novel method for rapid determination of carbonyl in lignins by headspace gas chromatography (HS-GC). The method involves the quantitative carbonyl reduction for aldehydes in 2min at room temperature or for acetones in 30min at 80°C
Determination of mercury in natural waters and effluents by flameless atomic absorption spectrophotometry.
S H Omang
Analytica chimica acta, 53(2), 415-419 (1971-02-01)
Sina Abdolhosseinzadeh et al.
Scientific reports, 5, 10160-10160 (2015-05-16)
Exfoliation of graphite is a promising approach for large-scale production of graphene. Oxidation of graphite effectively facilitates the exfoliation process, yet necessitates several lengthy washing and reduction processes to convert the exfoliated graphite oxide (graphene oxide, GO) to reduced graphene

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service