Skip to Content
Merck
All Photos(1)

Key Documents

SML0124

Sigma-Aldrich

Rasagiline mesylate

≥98% (HPLC)

Synonym(s):

(1R)-2,3-Dihydro-N-2-propynl-1H-inden-1-amine methanesulfonate, N-Propargyl-1(R)-aminoindan methanesulfonate, TVP-1012

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N · CH4O3S
CAS Number:
Molecular Weight:
267.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +15 to +28°, c = 0.6 mg/mL in H2O

storage condition

desiccated

color

white to tan

solubility

H2O: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.C#CCN[C@@H]1CCc2ccccc12

InChI

1S/C12H13N.CH4O3S/c1-2-9-13-12-8-7-10-5-3-4-6-11(10)12;1-5(2,3)4/h1,3-6,12-13H,7-9H2;1H3,(H,2,3,4)/t12-;/m1./s1

InChI key

JDBJJCWRXSVHOQ-UTONKHPSSA-N

Gene Information

human ... MAOB(4129)

Application

Rasagiline mesylate has been used:
  • to test its neuroprotective effects in retinitis pigmentosa (RP) by apoptosis regulator, Bax/Bcl-2 modulation
  • as monoamine oxidase-B inhibitor in 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) induced mouse model
  • as an anti-parkinsonian agent to test its rescue functionality in 6-hydroxydopamine-lesioned zebrafish larvae towards bradykinetic and dyskinetic-like behavior

Rasagiline mesylate may be used in MAO type B-mediated cell signaling studies.

Biochem/physiol Actions

Anti-Parkinson drug; MAO B Inhibitor; anti-apoptotic; neuroprotectant
Rasagiline mesylate is an effective therapeutic option in early stages of Parkinson′s disease. It improves the motor fluctuations in levodopa-treated patients and may be a useful adjunct to such patients. It has neuroprotective effects and increases the survival of dopaminergic neurons.
Rasagiline mesylate is an irreversible inhibitor of monoamine oxidase selective for MAO type B over type A by a factor of fourteen. It has anti-apoptotic and neuroprotectant activity and has been used as a treatment for Parkinson′s disease.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism and Histamine Synthesis and Metabolism pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoka Kasai et al.
Frontiers in behavioral neuroscience, 11, 75-75 (2017-05-19)
Parkinson's disease (PD), a neurodegenerative disorder, is accompanied by various non-motor symptoms including depression and anxiety, which may precede the onset of motor symptoms. Selegiline is an irreversible monoamine oxidase-B (MAO-B) inhibitor, and is widely used in the treatment of
Dannit Licht et al.
The Journal of pharmacy and pharmacology, 68(10), 1278-1289 (2016-07-29)
While bioequivalence between enteric-coated and immediate-release formulations can be achieved in terms of AUC, gastric emptying of enteric-coated dosage forms is a stochastic event, usually leading to lower Cmax values than those observed with the corresponding immediate release. This article
O Rascol et al.
Lancet (London, England), 365(9463), 947-954 (2005-03-16)
Rasagiline mesylate is a novel drug for Parkinson's disease with selective, irreversible monoamine oxidase B (MAO-B) inhibitor activity, and is effective as monotherapy in early disease. This study investigated rasagiline efficacy and safety in levodopa-treated patients with Parkinson's disease and
J M Rabey et al.
Clinical neuropharmacology, 23(6), 324-330 (2001-09-29)
Rasagiline mesylate (TVP-1012) is a potent, selective, non-reversible MAO-B inhibitor, without the tyramine-potentiating effect and with neuroprotective activities. The benefit of rasagiline as monotherapy in patients with early Parkinson's disease (PD) has already been reported. To evaluate the safety, tolerability
Rita L Vaz et al.
Pharmacology, biochemistry, and behavior, 189, 172828-172828 (2019-12-01)
Parkinson's disease (PD) is known as a movement disorder due to characteristic motor features. Existing therapies for PD are only symptomatic, and their efficacy decreases as disease progresses. Zebrafish, a vertebrate in which parkinsonism has been modelled, offers unique features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service