Skip to Content
Merck
All Photos(1)

Key Documents

SML0150

Sigma-Aldrich

Tolcapone

≥98% (HPLC)

Synonym(s):

3,4-Dihydroxy-4′-methyl-5-nitrobenzophenone, Ro 40-7592

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11NO5
CAS Number:
Molecular Weight:
273.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow

solubility

DMSO: ≥15 mg/mL

originator

Roche

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)C(=O)c2cc(O)c(O)c(c2)[N+]([O-])=O

InChI

1S/C14H11NO5/c1-8-2-4-9(5-3-8)13(17)10-6-11(15(19)20)14(18)12(16)7-10/h2-7,16,18H,1H3

InChI key

MIQPIUSUKVNLNT-UHFFFAOYSA-N

Gene Information

human ... COMT(1312)

Looking for similar products? Visit Product Comparison Guide

Application

Tolcapone may be used in COMT-mediated cell signaling studies.
Tolcapone has been used in methyltransferase assay in human embryonic kidney 293 cells.

Biochem/physiol Actions

Inhibition of catechol-O-methyltransferase by tolcapone increases the half-life and bioavailabilty of levodopa. It therefore is an effective adjunctive therapy in patients with Parkinson′s disease with motor response fluctuations.
Tolcapone is an orally active catechol-O-methyltransferase (COMT) inhibitor. It inhibits both central and peripheral COMT. Tolcapone crosses the blood-brain barrier, and has been used for L-DOPA adjunct therapy in the treatment of Parkinson′s Disease.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenny J Fischer et al.
Toxicological sciences : an official journal of the Society of Toxicology, 113(1), 243-253 (2009-09-29)
Capture compound mass spectrometry (CCMS) is a novel technology that helps understand the molecular mechanism of the mode of action of small molecules. The Capture Compounds are trifunctional probes: A selectivity function (the drug) interacts with the proteins in a
H Baas et al.
Journal of neurology, neurosurgery, and psychiatry, 63(4), 421-428 (1997-10-29)
More than 50% of patients with Parkinson's disease develop motor response fluctuations (the "wearing off" phenomenon) after more than five years of levodopa therapy. Inhibition of catechol-O-methyltransferase by tolcapone has been shown to increase levodopa bioavailability and plasma elimination half
Gillian M Keating et al.
CNS drugs, 19(2), 165-184 (2005-02-09)
Tolcapone (Tasmar) is a selective, reversible inhibitor of peripheral and central catechol-O-methyltransferase (COMT). Results of well designed studies indicate that oral tolcapone is an effective adjunct to levodopa plus a peripheral dopa-decarboxylase inhibitor (DDCI) in patients with fluctuating Parkinson's disease.
Daniel D Truong
Clinical interventions in aging, 4, 109-113 (2009-06-09)
Levodopa has been the gold standard therapy for the motor symptoms of Parkinson's disease for more than three decades. Although it remains the most effective treatment, its long-term use is associated with motor fluctuations and dyskinesias that can be disabling
Development of an HTRF assay for the detection and characterization of inhibitors of catechol-O-methyltransferase
Kimos M, et al.
Journal of Biomolecular Screening, 21, 490-495 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service