Skip to Content
Merck

Skip To

B7283

Benserazide hydrochloride

≥98% (HPLC), solid, L-aromatic amino acid decarboxylase inhibitor

Synonym(s):

DL-Serine 2-(2,3,4-trihydroxybenzyl)hydrazide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H15N3O5 · HCl
CAS Number:
Molecular Weight:
293.70
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352125
EC Number:
238-991-9
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Benserazide hydrochloride, ≥98% (HPLC), solid

biological source

synthetic

Quality Level

assay

≥98% (HPLC)

form

solid

solubility

H2O: 10 mg/mL

originator

Roche

storage temp.

room temp

SMILES string

Cl.NC(CO)C(=O)NNCc1ccc(O)c(O)c1O

InChI

1S/C10H15N3O5.ClH/c11-6(4-14)10(18)13-12-3-5-1-2-7(15)9(17)8(5)16;/h1-2,6,12,14-17H,3-4,11H2,(H,13,18);1H

InChI key

ULFCBIUXQQYDEI-UHFFFAOYSA-N

Gene Information

human ... DDC(1644)

General description

Benserazide is a peripheral decarboxylase inhibitor.

Application

Benserazide has been used:
  • to inhibit dopa-decarboxylase
  • to induce dyskinesia
  • to test on purified bacterial TDCs to assess the extent to which it could affect bacterial decarboxylases
  • as a peripheral dopa decarboxylase inhibitor to treat mice

Biochem/physiol Actions

Inhibitor of L-aromatic amino acid decarboxylase.
Benserazide can block HK2 enzymatic activity in vitro. It is used along with levodopa to treat Parkinson′s disease.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B0477000Y0002115BP685
form

solid

form

-

form

-

form

solid

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

storage temp.

room temp

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: 10 mg/mL

solubility

-

solubility

-

solubility

-

originator

Roche

originator

-

originator

-

originator

-


Still not finding the right product?

Explore all of our products under Benserazide hydrochloride


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Developmental and evolutionary basis for drought tolerance of the Anopheles gambiae embryo
Goltsev Y, et al.
Developmental Biology, 330(2), 462-470 (2009)
24 - Extrapyramidal movement disorders and spasticity
Waller DG, et al.
Medical Microbiology, 325-336 (2018)
Restoration of the dopamine transporter through cell therapy improves dyskinesia in a rat model of Parkinson?s Disease
Tomas D, et al.
PLoS ONE, 11(4), e0153424-e0153424 (2016)



Global Trade Item Number

SKUGTIN
B7283-1G04061833441718
B7283-5G04061833441725

Questions

  1. What is the recommended storage temperature for Benserazide hydrochloride (Product B7283) after the drug is suspended in water?

    1 answer
    1. Benserazide hydrochloride is soluble in water and should be dissolved in oxygen-free boiled water containing 0.1% sodium metabisulfite or other antioxidants. The solutions decompose on exposure to light and air. Benserazide HCl is a tan, photosensitive solid and should be stored tightly sealed at -4°C, protected from exposure to light. It is not recommended to store the aqueous solution for more than one day.

      Additionally, the product is subject to oxidation, and it is advisable to make solutions fresh daily. The product should be stored, as supplied, at room temperature for up to 1 year, and it should be protected from light.

      Helpful?

Reviews

No rating value

Active Filters