Skip to Content
Merck
All Photos(1)

Documents

325341

Sigma-Aldrich

Propargyl benzenesulfonate

96%

Synonym(s):

2-Propyn-1-yl benzenesulfonate, 2-Propynyl benzenesulfonate, Propargyl besylate, Propargyl phenylsulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO3CH2C≡CH
CAS Number:
Molecular Weight:
196.22
Beilstein:
1107228
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.525 (lit.)

bp

140-142 °C/2 mmHg (lit.)

mp

−30 °C (lit.)

density

1.243 g/mL at 25 °C (lit.)

SMILES string

O=S(=O)(OCC#C)c1ccccc1

InChI

1S/C9H8O3S/c1-2-8-12-13(10,11)9-6-4-3-5-7-9/h1,3-7H,8H2

InChI key

RAGBYXLIHQFIPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service