Skip to Content
Merck
All Photos(1)

Key Documents

P-007

Supelco

PCP (Phencyclidine) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Phencyclidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25N
CAS Number:
Molecular Weight:
243.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

N3(CCCCC3)C2(CCCCC2)c1ccccc1

InChI

1S/C17H25N/c1-4-10-16(11-5-1)17(12-6-2-7-13-17)18-14-8-3-9-15-18/h1,4-5,10-11H,2-3,6-9,12-15H2

InChI key

JTJMJGYZQZDUJJ-UHFFFAOYSA-N

General description

Phencyclidine, commonly known as PCP, is a recreational drug with hallucinogenic and neurotoxic effects. With street names such as "angel dust" or "wet", PCP comes in either powder or liquid forms. The illicit drug is typically sprayed onto leafy material such as cannabis and then smoked. This certified solution standard is suitable for use as starting material in calibrators and controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masakuni Horiguchi et al.
Behavioural brain research, 238, 36-43 (2012-09-29)
Development of dopamine (DA) D(1) receptor agonists is a priority to improve cognitive impairment in schizophrenia (CIS). This study examined the dose-response relationship of the selective D(1) agonist, SKF38393 (0.5-40 mg/kg), to reverse the deficit in novel object recognition (NOR)
Ross Zimnisky et al.
Psychopharmacology, 226(1), 91-100 (2012-10-20)
A major challenge in the pharmacological treatment of psychotic disorders is the effective management of the associated cognitive dysfunctions. Novel concepts emphasize a potential benefit of partial agonists acting upon dopamine D(2)-like receptors in ameliorating these cognitive deficits, and pre-clinical
Ramin Hajikhani et al.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University, 21(3), 307-312 (2012-12-12)
The study attempted to investigate the anti-anxiety activities of Phencyclidine (1-(1-phenylcyclohexyl) piperidine, PCP, I) and some of its derivatives (M, F, L, B, S, P) with the elevated-plus maze (EPM) Test. Phencyclidine and its derivatives (M, F, L, B, S
Jens Köhler et al.
Journal of medicinal chemistry, 55(20), 8953-8957 (2012-09-28)
We synthesized and investigated the NMDA and σ₁ receptor affinity of enantiomerically pure 2-(2-phenyl-1,3-dioxan-4-yl)ethanamines 17-26. The primary amines (R,R)-18-20 with an axially oriented phenyl moiety in position 2 interacted with high enantioselectivity (eudismic ratios 70-130) and high affinity (K(i)((R,R)-19) =
Daniel C Javitt et al.
Schizophrenia bulletin, 38(5), 958-966 (2012-09-19)
Over the last 20 years, glutamatergic models of schizophrenia have become increasingly accepted as etiopathological models of schizophrenia, based on the observation that phencyclidine (PCP) induces a schizophrenia-like psychosis by blocking neurotransmission at N-methyl-D-aspartate (NMDA)-type glutamate receptors. This article reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service