Skip to Content
Merck
All Photos(3)

Documents

A9978

Sigma-Aldrich

AICAR

≥98% (HPLC), powder, AMPK activator

Synonym(s):

5-Aminoimidazole-4-carboxamide 1-β-D-ribofuranoside, Acadesine, N1-(β-D-Ribofuranosyl)-5-aminoimidazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N4O5
CAS Number:
Molecular Weight:
258.23
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

AICAR, ≥98% (HPLC), powder

Assay

≥98% (HPLC)

form

powder

color

tan

mp

214-215 °C

solubility

H2O: >7 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

NC(=O)c1ncn([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c1N

InChI

1S/C9H14N4O5/c10-7-4(8(11)17)12-2-13(7)9-6(16)5(15)3(1-14)18-9/h2-3,5-6,9,14-16H,1,10H2,(H2,11,17)/t3-,5-,6-,9-/m1/s1

InChI key

RTRQQBHATOEIAF-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adenosine monophosphate protein kinase (AMPK) activator 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR) modulates cellular energy. It regulates lipid and glucose metabolism, pro-inflammatory responses, cytokine production, cell proliferation and apoptosis. AICAR improves ischemia or reperfusion injury and kidney fibrosis in rats. It provides protection against acute tubular necrosis.

Application

AICAR has been used to provide protection against cisplatin-induced acute kidney injury through the JAK (tyrosine-protein kinase )/STAT (signal transducer and activator of transcription )/SOCS (suppressor of cytokine signaling) pathway. It also has been used in the assay of FapR (malonyl-CoA responsive bacterial transcriptional factor )-NLuc (nanoLuciferase) biosensor activity.

Biochem/physiol Actions

AICAR is a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes. In vivo, pharmacologic activation of AMPK with AICAR mimics exercise and triggers insulin-independent glucose uptake by skeletal muscle.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ai Ushiyama et al.
Biology of reproduction, 100(6), 1482-1491 (2019-02-06)
Despite knowledge that glucose metabolism is essential for the regulation of signaling cascades in the sperm that are pre-assembled into specific areas and function at multistage for fertilization, the physiological roles of glucose in avian sperm are poorly understood. Accumulated
Genetically-encoded FapR-NLuc as a biosensor to determine malonyl-CoA in situ at subcellular scales
Du Y, et al.
Bioconjugate Chemistry (2019)
Hiroaki Sunaga et al.
Scientific reports, 9(1), 11841-11841 (2019-08-16)
Fibroblast growth factor 21 (FGF21) is a metabolic hormone having anti-oxidative and anti-hypertrophic effects. However, the regulation of FGF21 expression during acute myocardial infarction (AMI) remains unclear. We tested blood samples from 50 patients with AMI and 43 patients with
Hongshun Wang et al.
Cell death & disease, 10(9), 627-627 (2019-08-21)
Oxysterol-binding protein like 2 (OSBPL2) was identified as a novel causal gene for autosomal dominant nonsyndromic hearing loss. However, the pathogenesis of OSBPL2 deficits in ADNSHL was still unclear. The function of OSBPL2 as a lipid-sensing regulator in multiple cellular
Sofija Mijic et al.
Nature communications, 8(1), 859-859 (2017-10-19)
Besides its role in homologous recombination, the tumor suppressor BRCA2 protects stalled replication forks from nucleolytic degradation. Defective fork stability contributes to chemotherapeutic sensitivity of BRCA2-defective tumors by yet-elusive mechanisms. Using DNA fiber spreading and direct visualization of replication intermediates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service