Skip to Content
Merck
All Photos(1)

Key Documents

P8013

Sigma-Aldrich

Pargyline hydrochloride

Synonym(s):

N-Methyl-N-(2-propynyl)benzylamine hydrochloride, N-Methyl-N-propargylbenzylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)CH2C≡CH·HCl
CAS Number:
Molecular Weight:
195.69
Beilstein:
3699487
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic

Assay

98% (TLC)

form

powder or crystals

mp

160-163 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Cl[H].CN(CC#C)Cc1ccccc1

InChI

1S/C11H13N.ClH/c1-3-9-12(2)10-11-7-5-4-6-8-11;/h1,4-8H,9-10H2,2H3;1H

InChI key

BCXCABRDBBWWGY-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Pargyline hydrochloride has been used:
  • as a histone demethylase inhibitor to treat monocytes to investigate whether various inhibitors affect training [1]
  • to maximize uptake into dopaminergic neurons (DA) neurons in experimental rats[2],
  • to study its effect on discolouration of medullary tissues of sclerotia of S. sclerotiorum[3]

Biochem/physiol Actions

Pargyline hydrochloride acts as a histone demethylase inhibitor.[1] It has antihypertensive action.[4]
Selective MAO-B inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemical Pathway for the Formation of Abnormal Sclerotia of Sclerotinia sclerotiorum
Huang HC and Yeung JM
Plant Pathology, 11(1), 2-2 (2002)
Trained immunity or tolerance: opposing functional programs induced in human monocytes after engagement of various pattern recognition receptors
Ifrim DC, et al.
Clinical and Vaccine Immunology : CVI, 21(4), 534-545 (2014)
An update on antidepressant toxicity: an evolution of unique toxicities to master
Liebelt EL
Clinical Pediatric Emergency Medicine, 9(1), 24-34 (2008)
Resonant antidromic cortical circuit activation as a consequence of high-frequency subthalamic deep-brain stimulation
Li S, et al.
Journal of Neurophysiology, 98(6), 3525-3537 (2007)
Erica P Cai et al.
Nature metabolism, 2(9), 934-945 (2020-07-29)
Type 1 diabetes (T1D) is caused by the autoimmune destruction of pancreatic beta cells. Pluripotent stem cells can now be differentiated into beta cells, thus raising the prospect of a cell replacement therapy for T1D. However, autoimmunity would rapidly destroy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service