Skip to Content
Merck
All Photos(2)

Key Documents

RAB0277

Sigma-Aldrich

Rat IL-1 β ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

Il-1 beta, Interleukin-1 beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/mL
standard curve range: 68.59-50000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Il1b(24494)

General description

The Rat IL-1 β ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat IL-1 β in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Rat IL-1 β

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pegah Dejban et al.
European journal of pharmacology, 887, 173579-173579 (2020-09-21)
Gastric ulcer is a prevalent disease with various etiologies, including non-steroidal anti-inflammatory drugs (NSAIDs), stress conditions, and alcohol, resulting in an inflammatory condition in the gastric mucosa. The aim of this study was to explore the protective effects of modafinil
Miao Liu et al.
Biochimica et biophysica acta. Molecular basis of disease, 1864(5 Pt B), 1923-1930 (2017-10-27)
Amylin is a pancreatic β-cell hormone co-secreted with insulin, plays a role in normal glucose homeostasis, and forms amyloid in the pancreatic islets of individuals with type-2 diabetes. Aggregated amylin is also found in blood and extra-pancreatic tissues, including myocardium.
Dan-Dan Zhang et al.
Neurotoxicity research, 40(4), 1057-1069 (2022-06-15)
Neuropathic pain (NP) is a common disorder among individuals worldwide, but there is still no effective treatment for NP. The EGFR pathway promotes NP nociceptive sensitization and represents a potential therapeutic target. Geniposide is abundant in natural plants and has
Tao Li et al.
International journal of clinical and experimental pathology, 7(12), 8401-8410 (2015-02-13)
Anti-inflammatory and anti-apoptotic effects of polydatin (PD) have been demonstrated in our previous studies. Recently, we have found that PD treatment can ameliorate burn-induced acute lung injury (ALI). In the present study, we hypothesized that PD may provide protective effect
Wei Li et al.
BMC complementary medicine and therapies, 21(1), 10-10 (2021-01-08)
Curcumin has been demonstrated to exert anti-oxidant, anti-fibrotic, anti-inflammatory, and anti-cancer activities. This study was conducted to observe the effect and inner mechanism of curcumin in rats with radiation-induced liver damage (RILD). Thirty SD rats were classified into Control, Radiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service