RAB0278
Rat IL-1 β ELISA Kit
for cell and tissue lysates
Synonym(s):
Il-1 beta, Interleukin-1 beta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell lysate
sample type tissue lysate
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/mL
standard curve range: 68.59-50000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Il1b(24494)
General description
The Rat IL-1 β ELSA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat IL-1β cell lysate and tissue lysate.
Immunogen
Recombinant Rat IL-1 β
Application
For research use only. Not for use in diagnostic procedures
Rat IL-1 Β ELISA Kit has been used to measure IL-1 Β levels .
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Rat IL-1 Β ELISA Kit has been used to measure IL-1 Β levels .
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Pro-inflammatory cytokine IL-1 Β (IL1B), which is highly expressed on Β-adrenoreceptor in obese adipose tissues, is implicated in induction of uncoupling protein 1 (UCP1) expression via extracellular signal-regulated kinase (ERK) activation in adipocytes. Macrophage autophagy inhibits the production of inflammasome-dependent IL-1Β and thereby it plays a vital role in reducing acute toxin-induced liver injury and death. In rats, IL-1Β enhanced neurogenesis and plasticity of carotid bodies (CB) by triggering ERK1/2 activation. IL-1Β plays a vital role in alteration of glomerular hemodynamics via stimulating the production of prostaglandin E2 (PGE2) in the mesangial cells.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Macrophage autophagy limits acute toxic liver injury in mice through down regulation of interleukin-1?.
Ilyas G
Journal of Hepatology null
Ashraf Al-Brakati et al.
International journal of nanomedicine, 16, 4335-4349 (2021-07-09)
Selenium nanoparticles (SeNPs) have recently gained much attention in nanomedicine applications owing to their unique biological properties. Biosynthesis of SeNPs using nutraceuticals as lycopene (LYC) maximizes their stability and bioactivities. In this context, this study aimed to elucidate the renoprotective
Proinflammatory cytokine interleukin-1? suppresses cold-induced thermogenesis in adipocytes.
Goto T
Cytokine null
Tormentic acid inhibits H2O2-induced oxidative stress and inflammation in rat vascular smooth muscle cells via inhibition of the NF-?B signaling pathway.
Wang YL
Molecular Medicine Reports null
Interleukin-1? promotes the neurogenesis of carotid bodies by stimulating the activation of ERK1/2.
Xue F
Respiratory Physiology & Neurobiology null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service