Skip to Content
Merck
All Photos(3)

Key Documents

365548

Sigma-Aldrich

Direct Red 80

Dye content 25 %

Synonym(s):

Sirius Red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H26N10Na6O21S6
CAS Number:
Molecular Weight:
1373.07
Colour Index Number:
35780
Beilstein:
8185575
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 25%

technique(s)

titration: suitable

color

dark brown

λmax

528 nm

ε (extinction coefficient)

≥12000 at 277-283 nm in H2O
≥15000 at 223-235 nm in H2O
≥32000 at 524-530 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S([O-])(=O)=O)\N=N\c3ccc(cc3)S([O-])(=O)=O)c(cc4cc(NC(=O)Nc5ccc6c(O)c(\N=N\c7ccc(cc7S([O-])(=O)=O)\N=N\c8ccc(cc8)S([O-])(=O)=O)c(cc6c5)S([O-])(=O)=O)ccc14)S([O-])(=O)=O

InChI

1S/C45H32N10O21S6.6Na/c56-43-33-13-5-27(17-23(33)19-39(81(71,72)73)41(43)54-52-35-15-7-29(21-37(35)79(65,66)67)50-48-25-1-9-31(10-2-25)77(59,60)61)46-45(58)47-28-6-14-34-24(18-28)20-40(82(74,75)76)42(44(34)57)55-53-36-16-8-30(22-38(36)80(68,69)70)51-49-26-3-11-32(12-4-26)78(62,63)64;;;;;;/h1-22,56-57H,(H2,46,47,58)(H,59,60,61)(H,62,63,64)(H,65,66,67)(H,68,69,70)(H,71,72,73)(H,74,75,76);;;;;;/q;6*+1/p-6/b50-48+,51-49+,54-52+,55-53+;;;;;;

InChI key

YIQKLZYTHXTDDT-IJPLDVMMSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Direct Red 80 (C.I. 35780) is an extremely hydrophilic, very large tetrakisazo hexasulfonated anionic dye that has a large, planar aromatic system.It is also known as Sirius red F3B, chlorantine fast red 7BLN or 5BRL, durazol brilliant red B, pontamine fast red 7BNL, and sirius fast red.

Application

Direct Red 80 has been used as an ingredient in the preparation of saturated picric acid for immunohistochemical detection and quantitation of collagen in liver tissues.Other applications include -
  • detection of amyloid
  • Sirius red-Van Gieson methods for quantitation of collagen using image analysis methods
  • counterstaining applications substituting eosin in alum hematoxylins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BMP2 regulates Osterix through Msx2 and Runx2 during osteoblast differentiation.
Matsubara T, et al.
The Journal of Biological Chemistry, 283(43), 29119-29125 (2008)
Degradation of a Textile Dye C. I. Direct Red 80 by
Electrochemical Processes
.
Lopes A, et al.
Portugaliae Electrochimica Acta , 279-294 (2004)
Sabatini M, et al.
Cartilage and Osteoarthritis (2004)
Christopher A Miller et al.
Circulation. Cardiovascular imaging, 6(3), 373-383 (2013-04-05)
Extracellular matrix expansion is a key element of ventricular remodeling and a potential therapeutic target. Cardiovascular magnetic resonance (CMR) T1-mapping techniques are increasingly used to evaluate myocardial extracellular volume (ECV); however, the most widely applied methods are without histological validation.
In vivo inhibition of rat stellate cell activation by soluble transforming growth factor beta type II receptor: a potential new therapy for hepatic fibrosis.
George J, et al.
Proceedings of the National Academy of Sciences of the USA, 96(22), 12719-12724 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service