Skip to Content
Merck
All Photos(1)

Key Documents

P4707

Sigma-Aldrich

Poly-ʟ-Lysine Hydrobromide

synthetic, liquid, 0.01%, suitable for cell culture

Synonym(s):

PLL Solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160406
NACRES:
NA.75

Product Name

Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture

biological source

synthetic

sterility

sterile-filtered

product line

BioReagent

form

liquid

mol wt

70—150 kDa

packaging

pkg of 50 mL

concentration

0.01%

technique(s)

cell culture | mammalian: suitable

surface coverage

4 μg/cm2

impurities

endotoxin, tested

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

N([C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)O)C(=O)[C@@H](N)CCCCN

InChI

1S/C18H38N6O4/c19-10-4-1-7-13(22)16(25)23-14(8-2-5-11-20)17(26)24-15(18(27)28)9-3-6-12-21/h13-15H,1-12,19-22H2,(H,23,25)(H,24,26)(H,27,28)/t13-,14-,15-/m0/s1

InChI key

WBSCNDJQPKSPII-KKUMJFAQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Application

  • Poly-L-lysine coated glass coverslips have been used for mounting samples for scanning electron microscopy.
  • Poly-L-lysine coated wells were used for trapping of PC-12 cells on electrodes.
  • Poly-L-lysine-coated borosilicate glass slides were used for the seeding of viral vector-transduced cells.
Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations. Lower molecular weight poly-L-lysine (30,000-70,000) is less viscuous in solution, but higher molecular weight versions provide more attachment sites per molecule.

Biochem/physiol Actions

Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. When it is absorbed to the cell culture surface, poly-L-lysine functions to increase the number of positively charged sites available for cell binding. With cells that can digest poly-L-lysine, poly-D-lysine should be used as the attachment factor.

Components

Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Preparation Note

This product is supplied as a 0.01% sterile filtered solution in water. Coating slides with this solution should be followed with a 5 minute incubation and drying the slides at room temperature. Coated slides will be stable for one year if protected from dust.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microparticles mediate MRP1 intercellular transfer and the re-templating of intrinsic resistance pathways.
Lu JF et al.
Pharmacological Research, 76, 77-77 (2013)
Promiscuous dimerization of the growth hormone secretagogue receptor (GHS-R1a) attenuates ghrelin-mediated signaling.
Schellekens H et al.
The Journal of Biological Chemistry, 288, 181-181 (2013)
All polymer chip for amperometric studies of transmitter release from large groups of neuronal cells.
Larsen ST and Taboryski R
Analyst, 137, 5057-5057 (2012)
Sahar Roshanbin et al.
BMC neuroscience, 17(1), 43-43 (2016-07-02)
MCT14 (SLC16A14) is an orphan member of the monocarboxylate transporter (MCT) family, also known as the SLC16 family of secondary active transmembrane transporters. Available expression data for this transporter is limited, and in this paper we aim to characterize MCT14
Alexandre Stipanovich et al.
Nature, 453(7197), 879-884 (2008-05-23)
Dopamine orchestrates motor behaviour and reward-driven learning. Perturbations of dopamine signalling have been implicated in several neurological and psychiatric disorders, and in drug addiction. The actions of dopamine are mediated in part by the regulation of gene expression in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service