Skip to Content
Merck
All Photos(2)

Documents

RAB0097

Sigma-Aldrich

Rat CRP/C-Reactive Protein ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

C-reactive protein, CRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 200 pg/mL
standard curve range: 0.246-60 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Crp(12944)

General description

Rat CRP/C-Reactive Protein ELISA (Enzyme-Linked Immunosorbent Assay) Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of a target protein in biological samples, such as serum, plasma, cell culture supernatants, urine, and/or cell and tissue lysates. C-reactive protein (CRP) belongs to the pentraxin family of proteins and is highly conserved. It is produced as a response to several inflammatory cytokines by the liver.

Immunogen

Recombinant Rat CRP

Application

Rat CRP/C-Reactive Protein ELISA Kit has been used for the quantification of C-reactive protein (CRP) in rat serum or rat acinar cell line (AR42J) and rat blood.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Intrinsically Disordered Motif Mediates Diverse Actions of Monomeric C-reactive Protein.
Li HY
The Journal of Biological Chemistry (2016)
Zahra Farahnak et al.
The Journal of nutrition, 149(3), 479-487 (2019-02-19)
Docosahexaenoic acid (DHA; 22:6n-3) is an n-3 (ω-3) fatty acid known for beneficial effects on body composition. The objective of the study was to test the dose response of lean and fat mass to DHA in healthy growing female rats.
Dose Responsive Effects of Subcutaneous Pentosan Polysulfate Injection in Mucopolysaccharidosis Type VI Rats and Comparison to Oral Treatment
Michael F
PLoS ONE (2014)
Curcumin protects the pancreas from acute pancreatitis via the mitogen-activated protein kinase signaling pathway
Wang Y, et al.
Molecular Medicine Reports, 20(4), 3027-3034 (2019)
Effects of C-Reactive Protein on the Cardiac Differentiation of Mouse Embryonic Stem Cells.
Jin Y
Biological & Pharmaceutical Bulletin (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service