RAB0246
Rat IL-10 ELISA Kit
for serum, plasma and cell culture supernatant
Synonym(s):
IL-10, Interleukin-10
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 8.23-6000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Il10(25325)
Related Categories
General description
The Rat IL-10 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of rat IL-10 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Rat IL-10
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Huanhuan Jiang et al.
Experimental and therapeutic medicine, 18(1), 459-466 (2019-07-02)
The aim of the present study was to explore the protective role of resveratrol (RES) in asthma-induced airway inflammation and remodeling, as well as its underlying mechanism. An asthma rat model was induced by ovalbumin (OVA) treatment. Rats were randomly
Qian Zhang et al.
PeerJ, 8, e10310-e10310 (2020-11-27)
Increasing evidence shows that maternal overnutrition may increase the risk of diabetes in offspring. We hypothesized that maternal sitagliptin intervention may improve glucose intolerance through gut targeting. Female Sprague-Dawley (SD) rats were fed a normal diet (ND) or a high-fat
Ting-Yu Chao et al.
Food science & nutrition, 9(9), 4758-4769 (2021-09-18)
Liver diseases, including viral hepatitis, liver cirrhosis, and liver cancer, mostly remain silent until the late stages and pose a continuing threat to millions of people worldwide. Liver transplantation is the most appropriate solution in the case of liver failure
Thomas B Sundberg et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(34), 12468-12473 (2014-08-13)
Genetic alterations that reduce the function of the immunoregulatory cytokine IL-10 contribute to colitis in mouse and man. Myeloid cells such as macrophages (MΦs) and dendritic cells (DCs) play an essential role in determining the relative abundance of IL-10 versus
Jyoti Joshi et al.
Parasitology international, 63(4), 612-620 (2014-04-22)
Visceral leishmaniasis (VL) represents the second most challenging infectious disease worldwide, leading to nearly 500,000 new cases and 60,000 deaths annually. Ninety per cent of VL cases occur in five countries namely Bangladesh, India, Nepal, Sudan and Brazil. No licensed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service