Skip to Content
Merck
All Photos(6)

Key Documents

P8250

Sigma-Aldrich

Peroxidase from horseradish

Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)

Synonym(s):

Donor:hydrogen-peroxide oxidoreductase, Horseradish peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

horseradish

Quality Level

type

Type II

form

essentially salt-free, lyophilized powder

specific activity

150-250 units/mg solid (using pyrogallol)

mol wt

~44 kDa

solubility

0.1 M phosphate buffer: soluble (pH 6.0)
H2O: soluble

absorbance ratio

RZ ≥1.8

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

SMILES string

[O+H2]O[O-]

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Horseradish peroxidase is isolated from horseradish roots (Amoracia rusticana) and belongs to the ferroprotoporphyrin group of peroxidases. HRP is a single chain polypeptide containing four disulfide bridges. It is a glycoprotein containing 18% carbohydrate. The carbohydrate composition consists of galactose, arabinose, xylose, fucose, mannose, mannosamine, and galactosamine depending upon the specific isozyme. Its molecular weight (~44 kDa) includes the polypeptide chain (33,890 Daltons), hemin plus Ca2+ (~700 Daltons), and carbohydrate (~9,400 Daltons). At least seven isozymes of HRP exist. The isoelectric point for horseradish peroxidase isozymes ranges from 3.0 - 9.0.

Application

Horseradish peroxidase (HRP) is isolated from horseradish roots (Amoracia rusticana). It is used in biochemistry applications such as western blots, ELISA and Immunohistochemistry. Horseradish peroxidase is used to amplify a weak signal and increase detectability of a target molecule, such as a protein. Horseradish peroxidase, product P8250, has been used to study nonoral antigens in inflamed gingiva[1] and Ebola virus glycoprotein toxicity[2].
The enzyme has been used as a comparison during the peroxidase assay of extract from mature tall fescue leaf blades.[3] It has also been used to measure H2O2 production.[4]

Biochem/physiol Actions

HRP readily combines with hydrogen peroxide (H2O2) and the resultant [HRP-H2O2] complex can oxidize a wide variety of hydrogen donors. The optimal pH is 6.0-6.5 and the enzyme is most stable in the pH range of 5.0-9.0. HRP can be conjugated to antibodies by several different methods including glutaraldehyde, periodate oxidation, through disulfide bonds, and also via amino and thiol directed cross-linkers. It is smaller and more stable than the enzyme labels β-galactosidase and alkaline phosphatase. Hence, it is the most desired label. Also, its glycosylation leads to lower non-specific binding.[5] It is also used for the determination of glucose[6] and peroxides[7] in solution. Sodium azide, cyanide, L-cystine, dichromate, ethylenethiourea, hydroxylamine, sulfide, vanadate, p-aminobenzoic acid, and Cd2+, Co2+, Cu2+, Fe3+, Mn2+, Ni2+, Pb2+ ions are found to inhibit the enzyme activity.[8]
When incubated with a substrate, horseradish peroxidase produces a coloured, fluorimetric, or luminescent derivative of the labeled molecule, allowing quantification. Horseradish peroxidase has been shown to slightly reduce the level of inhibition in a cydAB mutant.

Quality

Preliminary work shows it to contain at least five isoenzymes.

Unit Definition

One pyrogallol unit will form 1.0 mg purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.

Analysis Note

The RZ (Reinheitszahl) is the absorbance ratio A403/A275 determined at 0.5-1.0 mg/ml in deionized water. It is a measure of hemin content, not enzymatic activity. Even preparations with high RZ may have low enzymatic activity.

Other Notes

View more information on peroxidase at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W E Bullock et al.
The Journal of experimental medicine, 165(1), 195-210 (1987-01-01)
The principal host cell of H. capsulatum (Hc) is the M phi within which the pathogenic yeast phase of the fungus multiplies during active disease. The initial interaction between Hc yeasts and M phi therefore is a crucial step in
S M Mallison et al.
Infection and immunity, 56(4), 823-830 (1988-04-01)
In vitro experimentation indicates that periodontitis-associated bacteria contain potent polyclonal B-cell activators (PBA). We reasoned that if PBA were operative in vivo, plasma cells specific for nonoral antigens should be present in the inflamed gingival tissues, which are characterized by
Chuan Chiang-Ni et al.
Journal of microbiology, immunology, and infection = Wei mian yu gan ran za zhi, 49(6), 837-842 (2015-02-05)
The phenotypic heterogeneity of the human pathogen Streptococcus pyogenes [group A streptococcus (GAS)] is associated with bacterial virulence variation. During invasive GAS infection, mutations in the two-component regulatory system covR/covS leads to increases in hyaluronic acid capsule production, virulence genes
J W Macadam et al.
Plant physiology, 99(3), 872-878 (1992-07-01)
Cessation of cell expansion has been associated with cell wall cross-linking reactions catalyzed by peroxidase. This study utilized two genotypes of tall fescue (Festuca arundinacea Schreb.) that differ in length of the leaf elongation zone to investigate the relationship between
Nancy J Sullivan et al.
Journal of virology, 79(1), 547-553 (2004-12-15)
Ebola virus infection causes a highly lethal hemorrhagic fever syndrome associated with profound immunosuppression through its ability to induce widespread inflammation and cellular damage. Though GP, the viral envelope glycoprotein, mediates many of these effects, the molecular events that underlie

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service