Skip to Content
Merck
All Photos(1)

Key Documents

10814407001

Roche

Peroxidase (POD)

EIA grade, from horseradish

Synonym(s):

POD

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

horseradish

Quality Level

Assay

90% (HPLC)

form

lyophilized (salt-free)

specific activity

~225 U/mg lyophilizate, 25 °C (Guaiacol, H2O2)

packaging

pkg of 100 mg

manufacturer/tradename

Roche

concentration

10 g/L

technique(s)

activity assay: suitable

color

red to brown

optimum pH

6.0-6.5

solubility

water: soluble at 68 °F

suitability

suitable for molecular biology

UniProt accession no.

application(s)

detection
life science and biopharma

foreign activity

ATPase <0.00100%, none detected
AcP <0.00100%
Catalase <0.700%

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

General description

Donor: hydrogen-peroxide oxidoreductase
Peroxidase can be coupled to other proteins via its amino groups, as well as its carbohydrate moiety. The reconstituted solution can be used directly for conjugation without prior dialysis.

Application

Peroxidase (POD) has been used in the spermine oxidase enzyme activity assay.[1]

Sequence

Blocking substances
Preparations does not contain substances that block the amino groups, but in contrast to other enzymes POD has only 2 to 3 free amino groups which are not accessible; even by extended dialysis of POD, the number of NH2-groups/mol does not change.

POD
POD contains 1 molecule of protoheme IX as prosthetic group and 2 molecules of Ca2+.
POD exists in > 10 isoenzymic forms (some sources estimate approx. 40 isoenzymes).

Carbohydrate Content
12 to 14.5 mg/100 mg Iyophilizate

Free Amino Groups
Free amino groups: 2 to 3 mol/mol enzyme

Preparation Note

Activator: Ammonia, pyridine, imidazole, all at pH >7; detergents
Working concentration: A suggested working ratio, for immediate use in coupling procedures, is 2 mg POD to 4 mg lgG (molar ratio = 2:1, POD:IgG).
Specific activity: approximately 225U/mg lyophilizate (+25°C, guaiacol, H2O2) approximately 1,000 U/mg lyophilizate (+25°C, ABTS, H2O2, pH 5.0) approximately 3,500 U/mg lyophilizate (+25°C, tetramethylbenzidine, H2O2)
Amino groups: 2 - 3moles/mole enzyme
Carbohydrates: 12.0 - 14.5% (w/w)
Isoenzymes: ≥90% isoenzyme C (HPLC)
Purity: single peak (HPLC)
Purity number: 3.0 - 3.5 (A403/A275)

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew C Goodwin et al.
Methods in molecular biology (Clifton, N.J.), 720, 173-181 (2011-02-15)
Spermine oxidase (SMO), the most recently characterized polyamine metabolic enzyme, catalyzes the direct back-conversion of spermine to spermidine in an FAD-dependent reaction that also yields the byproducts hydrogen peroxide (H(2)O(2)) and 3-aminopropanal. These metabolites, particularly H(2)O(2), have been implicated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service