Skip to Content
Merck
All Photos(2)

Key Documents

D9143

Sigma-Aldrich

o-Dianisidine

peroxidase substrate, chromogenic, powder

Synonym(s):

3,3′-Dimethoxybenzidine, Fast Blue B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O2
CAS Number:
Molecular Weight:
244.29
Beilstein:
1879884
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

o-Dianisidine, peroxidase substrate

form

powder

solubility

H2O: slightly soluble
aqueous acid: soluble

SMILES string

COc1cc(ccc1N)-c2ccc(N)c(OC)c2

InChI

1S/C14H16N2O2/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2/h3-8H,15-16H2,1-2H3

InChI key

JRBJSXQPQWSCCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

o-Dianisidine has been used for neutrophil myeloperoxidase activity (MPO) assay. It has also been used for staining erythrocytes.
as a constituent in neutral benzidine staining of dried blood smears

Caution

Darkens in storage.

Warning

Probably carcinogenic.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

402.8 °F - closed cup

Flash Point(C)

206 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FIP200 is required for the cell-autonomous maintenance of fetal hematopoietic stem cells
Liu Fei, et al.
Blood, 116(23), 4806-4814 (2010)
Amnion epithelial cells promote lung repair via lipoxin A4
Tan JL, et al.
Stem Cells Translational Medicine, 6(4), 1085-1095 (2017)
RNAi-mediated silencing of TNF-alphas converting enzyme to down-regulate soluble TNF-alpha production for treatment of acute and chronic colitis
Song Y, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 239, 231-241 (2016)
Trim33/Tif1-gamma is essential for macrophage and neutrophil mobilisation to developmental or inflammatory cues
Demy DL, et al.
Journal of Cell Science, jcs-203471 (2017)
Ben Brankatschk et al.
PloS one, 6(7), e21771-e21771 (2011-07-15)
In this paper, we report that the PX domain-containing protein SNX16, a member of the sorting nexin family, is associated with late endosome membranes. We find that SNX16 is selectively enriched on tubulo-cisternal elements of this membrane system, whose highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service