Skip to Content
Merck

Skip To

S8409

Superoxide Dismutase from bovine liver

ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)

Synonym(s):

SOD, Superoxide: superoxide oxidoreductase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-943-0
MDL number:
EC Number:
Specific activity:
2,000-6,000 units/mg protein (biuret)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

ammonium sulfate suspension

Quality Segment

specific activity

2,000-6,000 units/mg protein (biuret)

mol wt

32.5 kDa

UniProt accession no.

storage temp.

2-8°C

Gene Information

cow ... SOD1(281495)

General description

Research area: Cell Signaling

Superoxide dismutases are a group of low molecular weight metalloproteins present in all aerobic cells of plants, animals, and micro-organisms. They provide protection against damaging reactions with the superoxide radical anion (O2-) by catalyzing its disproportionation into oxygen and hydrogen peroxide.

Application

Superoxide Dismutase from bovine liver has been used:


  • for measuring mitochondrial hydrogen peroxide production in skeletal muscle
  • as a positive control in Cu(II) binding stability assay
  • as a positive control in superoxide dismutase assay
  • in a study to assess the inactivation of endothelial-derived relaxing factor by oxidized lipoproteins.
  • in a study to investigate the role of hydrogen peroxide in the cytotoxicity of the xanthine/xanthine oxidase system.

Biochem/physiol Actions

Superoxide Dismutase catalyzes the dismutation of superoxide radicals to hydrogen peroxide and molecular oxygen. It plays a critical role in the defense of cells against the toxic effects of oxygen radicals. It competes with nitric oxide (NO) for superoxide anion (which reacts with NO to form peroxynitrite), thereby SOD promotes the activity of NO. SOD has also been shown to suppress apoptosis in cultured rat ovarian follicles, neural cell lines, and transgenic mice.

Physical form

Suspension in 3.8 M (NH4)2SO4, pH 7.0

Analysis Note

For assay method, see McCord, J.M. and Fridovich,I., J. Biol. Chem., 244, 6049 (1969).

Other Notes

One unit will inhibit reduction of cytochrome c by 50% in a coupled system with xanthine oxidase at pH 7.8 at 25 °C in a 3.0 mL reaction volume. Xanthine oxidase concentration should produce an initial ΔA550 of 0.025 ± 0.005 per min.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S8160S7446G2501
description

ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)

description

lyophilized powder, ≥1500 units/mg protein

description

BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)

description

Type I, ammonium sulfate suspension, ≥40 units/mg protein

specific activity

2,000-6,000 units/mg protein (biuret)

specific activity

≥1500 units/mg protein

specific activity

≥4,500 units/mg protein

specific activity

≥40 units/mg protein

Gene Information

cow ... SOD1(281495)

Gene Information

cow ... SOD1(281495)

Gene Information

-

Gene Information

cow ... GLUD1(281785)

form

ammonium sulfate suspension

form

lyophilized powder

form

lyophilized powder

form

ammonium sulfate suspension

mol wt

32.5 kDa

mol wt

32.5 kDa

mol wt

32.5 kDa

mol wt

310-350 kDa

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

UniProt accession no.

P00442

UniProt accession no.

P00442

UniProt accession no.

-

UniProt accession no.

P00366


Still not finding the right product?


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters