A8511
Luminol
≥97% (HPLC)
Synonym(s):
3-Aminophthalhydrazide, 5-Amino-2,3-dihydro-1,4-phthalazinedione
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
Beilstein:
383929
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
powder or chunks
mp
>300 °C (lit.)
solubility
DMSO: 50 mg/mL, clear to slightly hazy
SMILES string
Nc1cccc2C(=O)NNC(=O)c12
InChI
1S/C8H7N3O2/c9-5-3-1-2-4-6(5)8(13)11-10-7(4)12/h1-3H,9H2,(H,10,12)(H,11,13)
InChI key
HWYHZTIRURJOHG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Luminol is a chemiluminescent compound, which belongs to the class of cyclic acyl hydrazides. It exhibits chemiluminescence, when oxidized by oxidizing agents. Luminol may be associated with nausea, vomiting and mucosa irritation in eyes and skin.
Application
Luminol has been used in chemiluminescence assay.
Features and Benefits
Used for chemiluminescence analysis of, for example, metal cations, blood, and glucococorticoids.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Reference values of reactive oxygen species in seminal ejaculates using chemiluminescence assay
Agarwal A, et al.
Journal of Assisted Reproduction and Genetics, 32(12), 1721-1729 (2015)
Luminol-based chemiluminescent signals: clinical and non-clinical application and future uses
Khan P, et al.
Applied Biochemistry and Biotechnology, 173(2), 333-355 (2014)
Christophe A Marquette et al.
Analytical and bioanalytical chemistry, 385(3), 546-554 (2006-05-23)
This critical review discusses the results published between 2000 and 2005 on the development of analytical systems based on the luminol chemiluminescent and electrochemiluminescent reactions. An increasing number of non-specific detection systems based on the enhancing, inhibiting or catalysing effect
Feng-Ming Hsu et al.
Scientific reports, 5, 10291-10291 (2015-05-20)
Esophageal cancer patients with pathological complete response (pCR) to neoadjuvant chemoradiation (CRT) have favorable outcomes. Currently, there was no reliable biomarker predicting the response to CRT. Perioperative circulating mRNA may be associated with prognosis, but its application for predicting treatment
Fuan Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(50), 16030-16036 (2012-10-20)
The Cu(2+)-dependent ligation DNAzyme is implemented as a biocatalyst for the colorimetric or chemiluminescence detection of Cu(2+) ions, Hg(2+) ions, or cocaine. These sensing platforms are based on the structural tailoring of the sequence of the Cu(2+)-dependent ligation DNAzyme for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service