Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Peroxidase from horseradish
Synonym(s):
Donor:hydrogen-peroxide oxidoreductase, Horseradish peroxidase, Peroxidase from horseradish
CAS Number:
9003-99-0
EC Number:
232-668-6
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (9)
Biological Source
Physical Form
Quality Segment
Application
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
P8375
Peroxidase from horseradish,
Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Expand
View Pricing
P8250
Peroxidase from horseradish,
Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Expand
View Pricing
P6782
Peroxidase from horseradish,
Type VI-A, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol), 950-2000 units/mg solid (using ABTS)
Expand
View Pricing
77332
Peroxidase from horseradish,
lyophilized, powder, ~150 U/mg
Expand
View Pricing
P8125
Peroxidase from horseradish,
Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Expand
View Pricing
P2088
Peroxidase from horseradish,
Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
Expand
View Pricing
SRE0082
Peroxidase from horseradish,
Expand
View Pricing
P6140
Peroxidase from horseradish,
Type X, ammonium sulfate suspension
Expand
View Pricing
P8415
Peroxidase from horseradish,
Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Expand
View Pricing