Skip to Content
Merck
All Photos(4)

Key Documents

A4685

Sigma-Aldrich

Luminol sodium salt

luminescent, ≥98% (titration), powder and chunks

Synonym(s):

3-Aminophthalhydrazide, 5-Amino-2,3-dihydro-1,4-phthalazinedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H6N3O2Na
CAS Number:
Molecular Weight:
199.14
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

Luminol sodium salt,

Assay

≥98% (titration)

form

powder and chunks

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless to faint red

SMILES string

O=C1NN([Na])C(C2=CC=CC(N)=C21)=O

InChI

1S/C8H7N3O2.Na/c9-5-3-1-2-4-6(5)8(13)11-10-7(4)12;/h1-3H,(H4,9,10,11,12,13);/q;+1/p-1

InChI key

RVJVDCVIJCBUTH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Luminol sodium salt has been used:

  • as a component of MEBSS buffer in a bioluminescence assay to detect peroxidase activity in whole blood samples
  • to study protein interaction in whole-cell lysates by immunoprecipitation
  • for in vitro reactive oxygen species (ROS) quantification in CD45+ and CD45− cells

Biochem/physiol Actions

Luminol is a redox-sensitive chemiluminescent compound. It emits blue luminescence when the basic aqueous solution of luminol having oxygen atoms come in contact with oxidizing agents like potassium ferricyanide. It is highly stable and is applicable in various fields such as analytical chemistry, metallurgy, biochemistry, and forensics for detecting reactive species. Luminol has the potential to detect defects in neutrophils ex vivo during oxidative metabolism defects such as chronic granulomatous disease and myeloperoxidase deficiency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemiluminescence detection in analytical chemistry
Lancaster, J. Steven
Endeavour, 16, 194-194 (1992)
Yantu Zhang et al.
Journal of chromatography. A, 1154(1-2), 260-268 (2007-04-24)
A novel method was developed for the simultaneous determination of glucocorticoid residues such as triamcinolone (TR), prednisolone (PR), hydrocortisone (HC), cortisone (CO), methylprednisolone (MP), dexamethasone (DE) and triamcinolone acetonide (TA) by high-performance liquid chromatography (HPLC) coupled with chemiluminescence (CL) detection.
New Developments in Chemiluminescence Research
Gill, Steven, K.
Aldrichimica Acta, 16, 59-59 (1983)
Opioids prevent regeneration in adult mammals through inhibition of ROS production
Elodie L, et al.
Scientific Reports, 8(1) (2018)
Chemilunimescence of luminol: The chemcial reaction
White E H, et al.
ACS Chemical Biology, 86(5), 940-941 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service