Skip to Content
Merck
All Photos(1)

Documents

P6140

Sigma-Aldrich

Peroxidase from horseradish

Type X, ammonium sulfate suspension

Synonym(s):

Donor:hydrogen-peroxide oxidoreductase, Horseradish peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

biological source

horseradish

Quality Level

type

Type X

form

ammonium sulfate suspension

specific activity

≥225 units/mg protein (biuret, using pyrogallol)

mol wt

~44 kDa

concentration

≥1.0 mg/mL

absorbance ratio

RZ 2.5-3.5

storage temp.

2-8°C

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Horseradish peroxidase is isolated from horseradish roots (Amoracia rusticana) and belongs to the ferroprotoporphyrin group of peroxidases. HRP is a single chain polypeptide containing four disulfide bridges. It is a glycoprotein containing 18% carbohydrate. The carbohydrate composition consists of galactose, arabinose, xylose, fucose, mannose, mannosamine, and galactosamine depending upon the specific isozyme. Its molecular weight (~44 kDa) includes the polypeptide chain (33,890 Daltons), hemin plus Ca2+ (~700 Daltons), and carbohydrate (~9,400 Daltons). At least seven isozymes of HRP exist. The isoelectric point for horseradish peroxidase isozymes ranges from 3.0 - 9.0.

Application

Horseradish peroxidase (HRP) is isolated from horseradish roots (Amoracia rusticana). It is used in biochemistry applications such as western blots, ELISA and Immunohistochemistry. Horseradish peroxidase is used to amplify a weak signal and increase detectability of a target molecule, such as a protein. Product P6140 has been used to detect low density lipoprotein (LDL).
The enzyme from Sigma has been used while assessing the skin sensitization potential of pro-haptens. It has also been used to show that peroxidase (PO) activity and its heat stability correlate with the availability of free Ca2+ ions.

Biochem/physiol Actions

HRP readily combines with hydrogen peroxide (H2O2) and the resultant [HRP-H2O2] complex can oxidize a wide variety of hydrogen donors. The optimal pH is 6.0-6.5 and the enzyme is most stable in the pH range of 5.0-9.0. HRP can be conjugated to antibodies by several different methods that include the use of glutaraldehyde, periodate oxidation, disulfide bonds, and also via amino- and thiol-directed cross-linkers. It is smaller and more stable than the enzyme labels, β-galactosidase and alkaline phosphatase. Hence, it is the most desired label. Also, its glycosylation leads to lower non-specific binding. It is also used for the determination of glucose and peroxides in solution. Sodium azide, cyanide, L-cystine, dichromate, ethylenethiourea, hydroxylamine, sulfide, vanadate, p-aminobenzoic acid, and Cd2+, Co2+, Cu2+, Fe3+, Mn2+, Ni2+, Pb2+ ions are found to inhibit its enzyme activity.
When incubated with a substrate, horseradish peroxidase produces a coloured, fluorimetric, or luminescent derivative of the labeled molecule, allowing quantification. Horseradish peroxidase has been shown to slightly reduce the level of inhibition in a cydAB mutant.

Unit Definition

One pyrogallol unit will form 1.0 mg purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.

Physical form

Crystalline suspension in 3.2 M (NH4)2SO4 solution containing potassium phosphate buffer, pH 6.0

Preparation Note

Water may be used to dilute suspension if needed.

Analysis Note

Preliminary studies indicate the presence of two basic and no acidic isoenzymes
The RZ (Reinheitszahl) is the absorbance ratio A403/A275 determined at 0.5-1.0 mg/ml in deionized water. It is a measure of hemin content, not enzymatic activity. Even preparations with high RZ may have low enzymatic activity.

Other Notes

View more information on peroxidase at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Stuart Thompson et al.
Plants (Basel, Switzerland), 10(7) (2021-07-03)
The extensibility of synthetic polymers is routinely modulated by the addition of lower molecular weight spacing molecules known as plasticizers, and there is some evidence that water may have similar effects on plant cell walls. Furthermore, it appears that changes
N Santanam et al.
Journal of lipid research, 39(11), 2111-2118 (1998-11-04)
Estradiol has been documented to inhibit the oxidation of low density lipoprotein (LDL). We show that physiological concentrations of estradiol do not inhibit the oxidation of LDL by copper. LDL samples isolated from a) premenopausal and postmenopausal women and from
Christoph Plieth et al.
Plant signaling & behavior, 7(6), 650-660 (2012-05-15)
In this paper we demonstrate how peroxidase (PO) activities and their heat stability correlate with the availability of free Ca(2+) ions. Calcium ions work as a molecular switch for PO activity and exert a protective function, rendering POs heat stable.
Y P Chau et al.
Acta anatomica, 153(2), 135-144 (1995-01-01)
Vascular permeability in various rat sympathetic ganglia, including superior cervical ganglia, thoracic ganglia and the celiac-mesenteric ganglia (CMG) complex, was investigated by using lanthanum and horseradish peroxidase (HRP) as tracers with special attention to the neuronal and small granule-containing (SGC)
G M Cook et al.
Microbiology (Reading, England), 144 ( Pt 12), 3297-3308 (1999-01-12)
Escherichia coli produces an extracellular factor that inhibits the aerobic growth of Cyd- mutants, defective in the synthesis or assembly of the cytochrome bd-type quinol oxidase. This paper shows that such a factor is the iron-chelating siderophore enterochelin. Mutants in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service