L7275
LB Broth (Lennox)
Tablet microbial growth medium
Synonym(s):
Lennox broth , Luria Bertani Broth
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
grade
for molecular biology
Quality Level
sterility
non-sterile
form
tablet
technique(s)
microbiological culture: suitable
application(s)
food and beverages
microbiology
storage temp.
room temp
suitability
nonselective for Escherichia coli
nonselective for coliforms
General description
Dehydrated Luria-Bertani (LB) broth is called the Lennox LB, which has amino acids as carbon source.
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids and carbohydrates in a low-salt formulation.
Application
LB Broth (Lennox) has been used for culturing Escherichia coli and bacterial artificial chromosomes (BACs).
Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added, including those that require low-salt conditions, such as Zeocin®.
Features and Benefits
Lennox LB tablets provide:
- Convenient small package to eliminate weighing
- Fast preparation – no stirring required
Components
10 g/L Tryptone
5 g/L Yeast Extract
5 g/L NaCl
2.2 g/L inert binding agents
5 g/L Yeast Extract
5 g/L NaCl
2.2 g/L inert binding agents
Reconstitution
Add each 1.1g tablet in 48.3mL water (no stirring required). Autoclave for 20 minutes at 121C to sterilize. Allow to cool before making additions, such as antibiotics (if desired).
Legal Information
Zeocin is a registered trademark of Cayla Sarl
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of antibacterial surfaces by plasma grafting of zinc oxide based nanocomposites onto polypropylene
Journal of Colloid and Interface Science, 402, 320-326 (2013)
Experiments in Molecular Genetics null
Escherichia coli Physiology in Luria-Bertani Broth
Journal of Bacteriology, 189(23), 8746-8746 (2007)
Simultaneous quantification of active carbon-and nitrogen-fixing communities and estimation of fixation rates using fluorescence in situ hybridization and flow cytometry
Applied and Environmental Microbiology, 80(21), 6750-6759 (2014)
Fluorescence in situ hybridization of ductal lavage samples identifies malignant phenotypes from cytologically normal cells in women with breast cancer
Cancer Cytopathology, 111(3), 185-191 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service