Skip to Content
Merck
All Photos(2)

Key Documents

L2897

Sigma-Aldrich

LB Broth with agar (Lennox)

Highly-referenced microbial growth powder medium with Agar, low salt, suitable for salt-sensitive E. coli culture.

Synonym(s):

Lennox broth, Luria Bertani Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

for molecular biology

Quality Level

sterility

non-sterile

form

powder

composition

Agar, 15 g/L
NaCl, 5 g/L
Tryptone, 10 g/L
Yeast Extract, 5 g/L

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids and carbohydrates in a low-salt formulation.

Application

LB Broth with agar (Lennox) has been used for:

  • the transformation of competent E. coli cells
  • to cultivate Ecc-15 bacteria
  • embedding scaffold containing tissue derived from induced pluripotent stem cells (iPSCs)
  • transmission electron microscopy (TEM) and immunohistochemistry (IHC)
It is suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added, including those that require low-salt conditions, such as Zeocin®.

Features and Benefits

Lennox LB powder with agar provides:
  • A budget-friendly alternative to pre-poured plates
  • Easy scale-up using larger package sizes
  • Standard formulation

Preparation Note

1. Suspend 35 g in 1 L of distilled water.
2. Heat to boiling while stirring to dissolve all ingredients completely.
3. Autoclave for 15 minutes at 121°C.
To prepare Lennox L Agar: Add 1 g glucose and proceed with preparation instructions as above.To prepare the medium of Enquist and Sternberg: Aseptically add 10 ml sterile 1 M magnesium sulfate after autoclaving.

Legal Information

Zeocin is a registered trademark of Cayla Sarl

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural analysis of three-dimensional human neural tissue derived from induced pluripotent stem cells
Brooks PT, et al.
Journal of Separation Science, 6.0, 337-337 (2016)
Farbod Alimohammadi et al.
Langmuir : the ACS journal of surfaces and colloids, 34(24), 7192-7200 (2018-05-22)
Two-dimensional (2D) nanomaterials have attracted considerable attention in biomedical and environmental applications due to their antimicrobial activity. In the interest of investigating the primary antimicrobial mode-of-action of 2D nanomaterials, we studied the antimicrobial properties of MnO2 and MoS2, toward Gram-positive
Ultrastructural visualization of the Mesenchymal-to-Epithelial Transition during reprogramming of human fibroblasts to induced pluripotent stem cells
Hoffding, MK and Hyttel, P
Current Stem Cell Research & Therapy, 14.0, 39-53 (2015)
Patricia E Carreira et al.
Mobile DNA, 7, 21-21 (2016-11-16)
LINE-1 (L1) retrotransposons are a notable endogenous source of mutagenesis in mammals. Notably, cancer cells can support unusual L1 retrotransposition and L1-associated sequence rearrangement mechanisms following DNA damage. Recent reports suggest that L1 is mobile in epithelial tumours and neural
Eric P Caragata et al.
Applied and environmental microbiology, 86(11) (2020-03-30)
Given the continued high prevalence of mosquito-transmitted diseases, there is a clear need to develop novel disease and vector control strategies. Biopesticides of microbial origin represent a promising source of new approaches to target disease-transmitting mosquito populations. Here, we describe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service