Skip to Content
Merck
All Photos(1)

Key Documents

S1797

Sigma-Aldrich

SOC Medium

For use in transformation

Synonym(s):

Super Optimal broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

for molecular biology

Quality Level

sterility

0.2 μm filtered

form

liquid

composition

Dextrose, 3.603 g/L
KCl, 0.186 g/L
MgSO4, 4.8 g/L
Tryptone, 20 g/L
Yeast extract, 5 g/L

technique(s)

microbiological culture: suitable

application(s)

agriculture

storage temp.

2-8°C

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Super optimal medium with catabolic repressor (SOC) medium is a glucose-rich microbial growth medium used primarily in the recovery step of Escherichia coli competent cell transformations. The culture of E.coli cells in this nutrient-rich microbial broth increases the transformation efficiency of recombinant plasmids containing toxic protein genes by inhibiting the unintended activation of the lac promoter carried on the cloning vector. SOC medium contains peptides, amino acids, water-soluble vitamins, and glucose in a low-salt formulation.

Application

SOC Medium has been used for the transformation of DH5α competent E-coli cells. It has also been used to culture white colonies post-cloning for the identification of the recombinant protein of interest.

Features and Benefits

SOC liquid provides:
  • Filter-sterilized, ready-to-use format
  • Convenient package sizes
  • Standard formulation

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Zhao et al.
Applied and environmental microbiology, 75(24), 7624-7630 (2009-10-27)
Ampicillin-resistant (Amp(r)) Salmonella enterica isolates (n = 344) representing 32 serotypes isolated from retail meats from 2002 to 2006 were tested for susceptibility to 21 other antimicrobial agents and screened for the presence of five beta-lactamase gene families (bla(CMY), bla(TEM)
Qiao-Yang Sun et al.
Analytical biochemistry, 394(1), 144-146 (2009-07-23)
In this paper, we report a useful protocol for cloning toxic protein genes. Use of the SOC medium, which is a glucose-containing rich medium, significantly improved the transformation efficiency of a recombinant plasmid containing a toxic plant subtilase SaSBT1 cDNA.
Rebekah M Martin et al.
mSystems, 3(3) (2018-07-03)
Despite insights gained through experimental models, the set of bacterial genes important for human infection is unclear for many of our most threatening pathogens. Klebsiella pneumoniae is a leading cause of health care-associated infections (HAIs) and commonly colonizes hospitalized patients
Detection of Cryptosporidium parvum Oocysts on Fresh Produce Using DNA Aptamers
Iqbal A
PLoS ONE (2015)
Daniel C Volke et al.
Bio-protocol, 11(4), e3917-e3917 (2021-03-19)
Precise genome engineering has become a commonplace technique for metabolic engineering. Also, insertion, deletion and alteration of genes and other functional DNA sequences are essential for understanding and engineering cells. Several techniques have been developed to this end (e.g., CRISPR/Cas-assisted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service