Skip to Content
Merck
All Photos(2)

Key Documents

L3147

Sigma-Aldrich

LB Broth with agar (Miller)

Highly-referenced nutrient-rich microbial growth powder medium with Agar, suitable for regular E.coli culture.

Synonym(s):

Luria Agar, Luria Bertani agar, Miller’s LB agar, lysogeny broth agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
eCl@ss:
42040102
NACRES:
NA.85

grade

Molecular Biology
for molecular biology

sterility

non-sterile

form

powder

composition

Agar, 15 g/L
NaCl, 10 g/L
Tryptone, 10 g/L
Yeast Extract, 5 g/L

technique(s)

microbiological culture: suitable

pH

6.8-7.2(4% solution)

application(s)

food and beverages
microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Miller′s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates. The addition of agar provides a solid medium for microbial growth.

Application

LB Broth with agar (Miller) has been used for the generation of single colony for Pseudomonas putida KT2440, plating E. coli and for the generation of pure colonies of C14 and MC8M6 of bacterial strains.
Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added.

Features and Benefits

Miller′s LB powder with agar provides:
  • Easy scale-up using larger package sizes
  • A budget-friendly alternative to pre-poured plates
  • Standard formulation

Preparation Note

1. Suspend 40 g in 1L of distilled water.
2. Heat to boiling while stirring to dissolve.
3. Autoclave for 15 minutes at 121°C.
4. Cool to 50°C prior to dispensing into sterile petri dishes.
To prepare the medium of Luria and Burrows: Add 1 g glucose to medium and proceed with preparation instructions above.
To prepare the medium of Luria, Adams and Ting (also known as LC agar): Aseptically add 25 ml of sterile 0.1 M calcium chloride after autoclaving.

Reconstitution

Add 40g powder to 1L water. Heat to boiling while stirring to dissolve powder. Autoclave for 15 minutes at 121C to sterilize. Allow to cool slightly before making additions, such as antibiotics (if desired).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miller, J.H.
Experiments in Molecular Genetics, 433-433 (1972)
Difco Manual, 239-239 (1998)
Multiple microbial activities for volatile organic compounds reduction by biofiltration
Civilini M.
Journal of the Air & Waste Management Association (1995), 56(7), 922-930 (2006)
Batch solar disinfection inactivates oocysts of Cryptosporidium parvum and cysts of Giardia muris in drinking water
McGuigan KG, et al.
Journal of Applied Microbiology, 101(2), 453-463 (2006)
Gang Liu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(6), 7718-7732 (2020-04-16)
Liver inflammation is a common extraintestinal manifestation in inflammatory bowel disease (IBD), yet, the mechanisms driving gut-liver axis inflammation remain poorly understood. IBD leads to a breakdown in the integrity of the intestinal barrier causing an increase in portal and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service