Skip to Content
Merck
All Photos(1)

Documents

L3272

Sigma-Aldrich

LB Broth with agar (Luria low salt)

Powder microbial growth medium

Synonym(s):

Luria Agar Base (Miller’s Modification)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

for molecular biology

Quality Level

sterility

non-sterile

form

powder

composition

Agar, 15 g/L
NaCl, 0.5 g/L
Tryptone, 10 g/L
Yeast Extract, 5 g/L

technique(s)

microbiological culture: suitable

pH

6.8-7.2(3.05% solution)

application(s)

food and beverages
microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Luria′s low salt LB is a microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates. The addition of agar provides a solid medium for microbial growth.

Application

LB Broth with agar (Luria low salt) has been used:
  • for culturing E. coli DH10B cells.
  • in the isolation of diazotrophic bacteria.
  • for transformation of competent E. coli strain Stbl3 for cloning.
Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added, including those that require low-salt conditions, such as Zeocin®.

Features and Benefits

Luria′s low salt LB powder with agar provides:
  • Easy scale-up using larger package sizes
  • A budget-friendly alternative to pre-poured plates
  • Standard formulation

Preparation Note

1. Suspend 30.5 g in 1 L of distilled water.
2. Heat to boiling while stirring to dissolve.
3. Autoclave for 15 minutes at 121°C.
4. Cool to 50°C prior to dispensing into petri dishes.

Legal Information

Zeocin is a registered trademark of Cayla Sarl

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased Expression of a Reporter Plasmid by Crotalid Venom Fraction
Islas AA, et al.
Journal of Animal and Veterinary Advances, 9(17), 2214-2219 (2010)
Difco Manual, 271-271 (1998)
Using plant growth-promoting rhizobacteria (PGPR) to improve plant growth
Grobelak A, et al.
Ecological Engineering, 84, 22-28 (2015)
Vidya Padmanabhan Nair et al.
STAR protocols, 3(2), 101281-101281 (2022-04-26)
Human endogenous retroviruses (HERVs) comprise many regulatory elements and can regulate host gene activity at different expression levels via multiple mechanisms. Here, we introduce a step-by-step protocol to activate or repress transcription of HERV-K(HML-2) elements using the CRISPRa and CRISPRi
Miller, J.H.
Experiments in Molecular Genetics, 433-433 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service