Skip to Content
Merck
All Photos(1)

Key Documents

L3522

Sigma-Aldrich

LB Broth (Miller)

Highly-referenced nutrient-rich microbial growth powder medium, suitable for regular E.coli culture

Synonym(s):

Luria Broth, Miller’s LB Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

Molecular Biology
for molecular biology

Quality Level

description

Miller′s Modification, 10 g/L NaCl

sterility

non-sterile

form

powder

composition

NaCl, 10 g/L
Tryptone, 10 g/L
Yeast Extract, 5 g/L

pH

6.8-7.2(2.5% solution)

application(s)

food and beverages

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Miller′s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates.

Application

LB Broth (Miller) has been used as a culture medium for E coli, Pseudomonas putida KT2440, Vibrio parahaemolyticus, and soil samples for the isolation of Bacillus spp.
Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added.

Biochem/physiol Actions

Luria Broth is used for maintenance and propagation of Escherichia coli. The E. coli grow faster in Luria Broth because the tryptone and yeast supply essential growth factors that the E. coli would otherwise have to synthesize. Luria Broth also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.

Features and Benefits

Miller′s LB powder provides:
  • Easy scale-up using larger package sizes
  • A budget-friendly alternative to liquid
  • Standard formulation

Preparation Note

1. Suspend 25 g in 1 L of distilled water.
2. Autoclave for 15 minutes at 121 °C.
To prepare the medium of Luria, Adams and Ting (also known as LC broth: Aseptically add 25 ml of sterile 0.1 M calcium chloride after autoclaving.

Reconstitution

Stir to suspend 25g powder in 1L water. Autoclave for 15 minutes at 121C to sterilize. Allow to cool before making additions, such as antibiotics (if desired).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Demeng Tan et al.
Microorganisms, 8(3) (2020-03-12)
Phage therapy is a potential and promising avenue for controlling the emergence and spread of multidrug-resistant (MDR) Klebsiella pneumoniae, however, the rapid development of anti-phage resistance has been identified as an obstacle to the development of phage therapy. Little is
Single-cell bacteria growth monitoring by automated DEP-facilitated image analysis
Peitz I and van Leeuwen R
Lab on a chip, 10(21), 2944-2951 (2010)
Selecting lipopeptide-producing, Fusarium-suppressing Bacillus spp.: Metabolomic and genomic probing of Bacillus velezensis NWUMFkBS10. 5
Adeniji A, et al.
MicrobiologyOpen, e742-e742 (2018)
Ikhlaas M Kasli et al.
AMB Express, 9(1), 5-5 (2019-01-09)
Production of recombinant proteins such as antibody fragments in the periplasm of the bacterium Escherichia coli has a number of advantages, including the ability to form disulphide bonds, aiding correct folding, and the relative ease of release and subsequent capture
Philippe M Fortune et al.
American journal of botany, 95(4), 454-464 (2008-04-01)
The origin of polyploid Bromus species of section Genea was investigated using molecular data. This group of annual species native from the Old-World is composed of three diploids, two tetraploids, one hexaploid, and one octoploid. Molecular cloning, sequencing, and phylogenetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service