Skip to Content
Merck
All Photos(2)

Documents

B5285

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt

≥98%, powder

Synonym(s):

BC-Indicator, X-GlcA, X-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12BrClNNaO7
CAS Number:
Molecular Weight:
444.59
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, ≥98%

Assay

≥98%

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C([O-])=O)Oc2c[nH]c3ccc(Br)c(Cl)c23

InChI

1S/C14H13BrClNO7.Na/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;/h1-3,9-12,14,17-20H,(H,21,22);/q;+1/p-1/t9-,10-,11+,12-,14+;/m0./s1

InChI key

IBLSVGDGSKUDCT-CYRSAHDMSA-M

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt has been used:
  • as a component in β-glucuronidase (GUS) assay buffer for histochemical GUS assay
  • to prepare X-Gluc or GUS staining solution
  • to confirm GUS expression in samples by X-gluc staining

Chromogenic substrate for β-glucuronidase (GUS) gene detection.

Biochem/physiol Actions

5-bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt (X-GlcA), a colorless substrate for β-glucuronidase enzyme, generates a blue color at the end of the reaction.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao Liu et al.
Bio-protocol, 7(14), e2446-e2446 (2017-07-20)
Determination of a gene expression in guard cells is essential for studying stomatal movements. GUS staining is one means of detecting the localization of a gene expression in guard cells. If a gene is specially expressed in guard cells, the
GUS Staining of Guard Cells to Identify Localised Guard Cell Gene Expression
Liu Z, et al.
Plant Physiology (2017)
Development of an efficient bi-directional promoter with tripartite enhancer employing three viral promoters
Patro S, et al.
Journal of Biotechnology, 163(3), 311- 317 (2013)
Agrobacterium tumefaciens-mediated Transformation of Walnut (Juglans regia)
Walawage SL, et al.
Plant Physiology (2014)
Grégory Montiel et al.
Plants (Basel, Switzerland), 9(4) (2020-04-08)
Until recently, the roles of plant MADS-box genes have mainly been characterized during inflorescence and flower differentiation. In order to precise the roles of AGAMOUS-LIKE 12, one of the few MADS-box genes preferentially expressed in roots, we placed its cDNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service