Skip to Content
Merck
All Photos(4)

Key Documents

M9130

Sigma-Aldrich

4-Methylumbelliferyl-β-D-glucuronide hydrate

≥98% (HPLC), BioReagent, for identification of transformed plants

Synonym(s):

4-Methylumbelliferyl-β-D-glucopyranosiduronic acid, MUG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O9 · xH2O
CAS Number:
Molecular Weight:
352.29 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.52

grade

Molecular Biology
for molecular biology

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

solubility

pyridine: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

O.CC1=CC(=O)Oc2cc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)ccc12

InChI

1S/C16H16O9.H2O/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22;/h2-5,11-14,16,18-20H,1H3,(H,21,22);1H2/t11-,12-,13+,14-,16+;/m0./s1

InChI key

URVSQZMOFUEQAW-YYHOVTOASA-N

Looking for similar products? Visit Product Comparison Guide

General description

MUG is a chemical commonly used for the detection of transformed plants. MUG shows low levels of fluorescence until treatment with glucuronidase (GUS) encoded in transformed plants. Transformed plants can be identified by their MUG fluorescence.

Application

Suitable for fluorescent detection of glucuronidase gene expression in plants.

Reconstitution

Product is soluble in molecular biology grade water at a concentration of 0.35 mg/ml.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L J Moberg
Applied and environmental microbiology, 50(6), 1383-1387 (1985-12-01)
An assay procedure to screen for Escherichia coli in foods by using 4-methylumbelliferyl-beta-D-glucuronide (MUG) incorporated into lauryl tryptose (LST) broth was evaluated. The beta-glucuronidase produced by E. coli cleaves the MUG substrate to yield a fluorescent end product. E. coli-negative
Andrew G Kunihiro et al.
Molecular nutrition & food research, 64(14), e2000072-e2000072 (2020-06-09)
Curcumin prevents bone loss in resorptive bone diseases and inhibits osteoclast formation, a key process driving bone loss. Curcumin circulates as an inactive glucuronide that can be deconjugated in situ by bone's high β-glucuronidase (GUSB) content, forming the active aglycone.
Xiaotian Zhong et al.
The Journal of biological chemistry, 295(10), 3115-3133 (2020-02-02)
The fortuitously discovered antiaging membrane protein αKlotho (Klotho) is highly expressed in the kidney, and deletion of the Klotho gene in mice causes a phenotype strikingly similar to that of chronic kidney disease (CKD). Klotho functions as a co-receptor for
Yuan Ruan et al.
Current biology : CB, 28(17), 2718-2729 (2018-08-28)
The capacity for sustained cell division within the plant meristem is a critical determinant of organ structure and performance. This capacity is diminished in mutants lacking the microtubule-associated protein CLASP and when brassinosteroid signaling is increased. Here, we discovered that
M C Thaller et al.
Journal of clinical microbiology, 26(4), 791-793 (1988-04-01)
A new selective, differential plating medium to screen the common gram-negative urinary tract pathogens is described. The medium combines adonitol fermentation, phenylalanine deaminase, and beta-glucuronidase tests and allows the indole and cytochrome oxidase tests to be performed directly from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service