Skip to Content
Merck
All Photos(1)

Documents

M5519

Sigma-Aldrich

Murashige and Skoog Basal Medium

powder, suitable for plant cell culture

Synonym(s):

MS0 Medium, MSO Medium, MS Basal Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.72

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Murashige and Skoog medium is a widely used plant tissue culture growth medium. M&S Basal Medium contains macronutrients that include high levels of nitrate and organic additives such as agar, sugars, vitamins and growth regulators. Important growth regulators frequently added to M&S include IAA (auxin/morphogen) and the Kinetin (cytokinin/cell division promoter).

Application

Murashige and Skoog medium has been used in plant tissue culture medium for plant transformation study.

Formula variant

With the macro- and micronutrients, and vitamins as described by Murashige and Skoog (1962).

Media Formulation

Quantity

Formulated to contain 4.4 grams of powder per liter of medium.

Preparation Note

Murashige and Skoog medium can be reconstituted from powder or by combining products that are major components of complete M&S medium, such as macronutrient mixtures and vitamin mixtures. Murashige and Skoog medium (M5519) contains the micronutrients and vitamins of the original classic formulation. It can be supplemented with sucrose, agar, auxins (IAA) and cytokinins (Kinetin) to generate a complete medium for growth plant tissue culture.

related product

Product No.
Description
Pricing

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joan E Krochko et al.
Methods in molecular biology (Clifton, N.J.), 773, 113-134 (2011-09-08)
In vitro assays for cytochrome P450 enzymes developed from plant-derived microsomal extracts have not been used extensively for the characterization and quantification of enzyme activities in plant tissues. We describe here an in vitro assay for abscisic acid (ABA) 8'-hydroxylase
Ajay Kumar et al.
BMC genomics, 13, 597-597 (2012-11-07)
Development of a high quality reference sequence is a daunting task in crops like wheat with large (~17Gb), highly repetitive (>80%) and polyploid genome. To achieve complete sequence assembly of such genomes, development of a high quality physical map is
M Guan et al.
Journal of experimental botany, 66(1), 203-212 (2014-10-16)
Nitrogen (N) remobilization from reserves to sinks is essential for seedling establishment and seed production. Cytosolic glutamine synthetase (GS1) is up-regulated during both seed germination and seed filling in plants. However, the specific roles of the individual GS1 isogenes with
Thierry G A Lonhienne et al.
Scientific reports, 4, 5055-5055 (2014-05-24)
Growth, morphogenesis and function of roots are influenced by the concentration and form of nutrients present in soils, including low molecular mass inorganic N (IN, ammonium, nitrate) and organic N (ON, e.g. amino acids). Proteins, ON of high molecular mass
Youjun Zhang et al.
Plant communications, 1(5), 100028-100028 (2020-12-29)
Although the use of stable transformation technology has led to great insight into gene function, its application in high-throughput studies remains arduous. Agro-infiltration have been widely used in species such as Nicotiana benthamiana for the rapid detection of gene expression and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service