Skip to Content
Merck
All Photos(2)

Key Documents

D134406

Sigma-Aldrich

3′,5′-Dimethoxy-4′-hydroxyacetophenone

97%

Synonym(s):

4′-Hydroxy-3′,5′-dimethoxyacetophenone, Acetosyringone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(OCH3)2COCH3
CAS Number:
Molecular Weight:
196.20
Beilstein:
1966119
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

124-127 °C (lit.)

SMILES string

COc1cc(cc(OC)c1O)C(C)=O

InChI

1S/C10H12O4/c1-6(11)7-4-8(13-2)10(12)9(5-7)14-3/h4-5,12H,1-3H3

InChI key

OJOBTAOGJIWAGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3′,5′-Dimethoxy-4′-hydroxyacetophenone has been used to activate the virulence (vir) genes of tumor-inducing (Ti) plasmid of Agrobacterium tumefaciens in Agrobacterium-mediated transformation (AMT) protocol.
3′,5′-Dimethoxy-4′-hydroxyacetophenone has been used to activate the virulence (vir) genes of tumor-inducing (Ti) plasmid of in Agrobacterium-mediated transformation (AMT) protocol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agrobacterium-mediated transformation of friable embryogenic calli and regeneration of transgenic cassava.
Bull SE, et al.
Nature Protocols, 4(12), 1845-1845 (2009)
Haipeng Li et al.
Plant methods, 14, 50-50 (2018-07-07)
Genetically modified cotton accounts for 64% of the world's cotton growing area (22.3 million hectares). The genome sequencing of the diploid cotton progenitors Gossypium raimondii and Gossypium arboreum as well as the cultivated Gossypium hirsutum has provided a wealth of
Tomoyuki Nabeshima et al.
Frontiers in plant science, 8, 1940-1940 (2017-12-19)
Chrysanthemum stunt viroid (CSVd) was inoculated into two chrysanthemum (
Johana C Misas Villamil et al.
Nature communications, 10(1), 1576-1576 (2019-04-07)
Ustilago maydis is a biotrophic fungus causing corn smut disease in maize. The secreted effector protein Pit2 is an inhibitor of papain-like cysteine proteases (PLCPs) essential for virulence. Pit2 inhibitory function relies on a conserved 14 amino acids motif (PID14).
Brendon Conlan et al.
Plant, cell & environment, 42(4), 1287-1301 (2018-10-31)
The folding and assembly of Rubisco large and small subunits into L8 S8 holoenzyme in chloroplasts involves many auxiliary factors, including the chaperone BSD2. Here we identify apparent intermediary Rubisco-BSD2 assembly complexes in the model C3 plant tobacco. We show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service