Skip to Content
Merck

Skip To

B4666

5-Bromo-4-chloro-3-indolyl α-D-N-acetylneuraminic acid sodium salt

≥90%, powder

Synonym(s):

X-NeuNAc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C19H21BrClN2O9Na
CAS Number:
Molecular Weight:
559.72
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

5-Bromo-4-chloro-3-indolyl α-D-N-acetylneuraminic acid sodium salt, ≥90%

Quality Level

assay

≥90%

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Na].CC(=O)NC1C(O)CC(OC1C(O)C(O)CO)(Oc2c[nH]c3ccc(Br)c(Cl)c23)C(O)=O

InChI

1S/C19H22BrClN2O9.Na.H/c1-7(25)23-15-10(26)4-19(18(29)30,32-17(15)16(28)11(27)6-24)31-12-5-22-9-3-2-8(20)14(21)13(9)12;;/h2-3,5,10-11,15-17,22,24,26-28H,4,6H2,1H3,(H,23,25)(H,29,30);;

InChI key

QOSVDASAURDXES-UHFFFAOYSA-N

General description

Chromogenic substrate for neuraminidase

Application

5-Bromo-4-chloro-3-indolyl α-D-N-acetylneuraminic acid sodium salt has been used as a substrate for determining the neuraminidase enzymatic activity of
  • avian mycoplasma samples
  • Ornithobacterium rhinotracheale, a poultry pathogen
  • canine mycoplasma samples


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B5285M8639N1516
assay

≥90%

assay

≥98%

assay

≥95% (HPLC)

assay

≥95%

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

solubility

water: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to light yellow

solubility

methanol: 25 mg/mL, clear, colorless to faintly yellow

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Nga T Lao et al.
PloS one, 4(10), e7354-e7354 (2009-10-10)
Translation is most often terminated when a ribosome encounters the first in-frame stop codon (UAA, UAG or UGA) in an mRNA. However, many viruses (and some cellular mRNAs) contain "stop" codons that cause a proportion of ribosomes to terminate and
A survey of avian Mycoplasma species for neuraminidase enzymatic activity
Bercic RL, et al.
Veterinary Microbiology, 130(3-4), 391-397 (2008)
Ornithobacterium rhinotracheale has neuraminidase activity causing desialylation of chicken and turkey serum and tracheal mucus glycoproteins
Kastelic S, et al.
Veterinary Microbiology, 162(2-4), 707-712 (2013)



Global Trade Item Number

SKUGTIN
B4666-10MG04061832921310
B4666-2MG04061832921327

Questions

Reviews

No rating value

Active Filters