GUSS
β-Glucuronidase Reporter Gene Staining Kit
Synonym(s):
β-Glucuronidase Staining Kit, Reporter Gene Kit, Staining Kit for Glucuronidase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.32
Recommended Products
usage
kit sufficient for 100 histochemical assays
Quality Level
technique(s)
microbe id | staining: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Kit is used to anaylze β glucuronidase (E.coli GUS gene) expression in transformed plants.
Application
The E. coli GUS gene is extensively used to analyze gene expression in transformed plants. Plants have low intrinsic GUS activity, and the E. coli gene is quite stable in plant cells. As a tag, GUS remains active at the N-terminus of fusion proteins.
Kit is suitable for biochemical activity assays, immunological assays and histochemical staining of tissue and cells.
Kit is suitable for biochemical activity assays, immunological assays and histochemical staining of tissue and cells.
Features and Benefits
- Histochemical staining of plant tissues expressing the E. coli GUS enzyme
- Ideally suited to plant expression studies due to very low GUS activity in plants and high enzyme stability
- GUS does not interfere with plant cell function or viability
Other Notes
The substrate used in this kit is X-GlcA (5-bromo-4-chloro-3-indolyl β-D-glucuronide) which results in an insoluble indigo-blue precipitate in transfected cells and tissues.
Kit Components Only
Product No.
Description
- 2× Fixation Buffer 25 mL
- 5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt, ≥98% 1 mL/vial
- Reagent A for gus staining kit 50 mL
- Reagent B for gus staining kit 200 μL
- Reagent C for gus staining kit 200 μL
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ken Naito et al.
Nature, 461(7267), 1130-1134 (2009-10-23)
High-copy-number transposable elements comprise the majority of eukaryotic genomes where they are major contributors to gene and genome evolution. However, it remains unclear how a host genome can survive a rapid burst of hundreds or thousands of insertions because such
Li-Ling Lin et al.
International journal of molecular sciences, 14(7), 14270-14286 (2013-07-11)
Brassinosteroids (BRs) are endogenous plant hormones and are essential for normal plant growth and development. MicroRNAs (miRNAs) of Arabidopsis thaliana are involved in mediating cell proliferation in leaves, stress tolerance, and root development. The specifics of BR mechanisms involving miRNAs
Kuo-Hsiang Chuang et al.
Bioconjugate chemistry, 17(3), 707-714 (2006-05-18)
Combination therapy can help overcome limitations in the treatment of heterogeneous tumors. In the current study, we examined whether multiple therapeutic agents could be targeted to anti-dansyl single-chain antibodies (DNS scFv) that were anchored on the plasma membrane of cancer
C-M Cheng et al.
Cancer gene therapy, 15(6), 393-401 (2008-03-29)
Increasing the specificity of chemotherapy may improve the efficacy of cancer treatment. Toward this aim, we developed a strain of bacteria to express enzymes for selective prodrug activation and non-invasive imaging in tumors. beta-glucuronidase and the luxCDABE gene cluster were
R A Jefferson et al.
The EMBO journal, 6(13), 3901-3907 (1987-12-20)
We have used the Escherichia coli beta-glucuronidase gene (GUS) as a gene fusion marker for analysis of gene expression in transformed plants. Higher plants tested lack intrinsic beta-glucuronidase activity, thus enhancing the sensitivity with which measurements can be made. We
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service