Skip to Content
Merck
All Photos(6)

Documents

N1627

Sigma-Aldrich

4-Nitrophenyl β-D-glucuronide

≥98% (TLC)

Synonym(s):

4-Nitrophenyl β-D-glucuronide, 4-Nitrophenyl-β-D-glucopyranosiduronic acid, PNPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO9
CAS Number:
Molecular Weight:
315.23
Beilstein:
1438576
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

Assay

≥98% (TLC)

form

powder

solubility

H2O: soluble 100 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2ccc(cc2)[N+]([O-])=O

InChI

1S/C12H13NO9/c14-7-8(15)10(11(17)18)22-12(9(7)16)21-6-3-1-5(2-4-6)13(19)20/h1-4,7-10,12,14-16H,(H,17,18)/t7-,8-,9+,10-,12+/m0/s1

InChI key

QSUILVWOWLUOEU-GOVZDWNOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl β-D-glucuronide has been used as substrate in acetate buffer to measure β-glucuronidase.

Substrates

Chromogenic substrate for β-glucuronidase (GUS) gene detection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E S Piruzian et al.
Molecular genetics and genomics : MGG, 266(5), 778-786 (2002-01-26)
The coding region of the licB gene from Clostridium thermocellum was truncated at the 3' end. The modified lichenase encoded by the construct (LicBM2) retained the most important properties of the enzyme - its high activity and thermostability. LicBM2 consists
A tool for monitoring Trichoderma harzianum: II. The use of a GUS transformant for ecological studies in the rhizosphere
Green Helge and Jensen D Funck
Phytopathology, 85(11), 1436-1440 (1995)
beta-N-Cyanoethyl acyl hydrazide derivatives: A new class of beta-glucuronidase inhibitors
Khan KM, et al.
Chemical & Pharmaceutical Bulletin, 50(11), 1443-1446 (2002)
Analysis of Fluorescent Compounds in Urine by Liquid Chromatography
T. Hanaia & J. Huberta
Journal of Liquid Chromatography, 7, 1627-1642 (1984)
H Iwabuchi et al.
Journal of chromatography, 414(1), 139-148 (1987-02-20)
Molecular-secondary-ion mass spectrometry (SIMS) is a suitable method for the analysis of nonvolatile substances such as conjugated metabolites of drugs. We have developed a simple method for the direct SIMS measurement of conjugates following thin-layer chromatography without any extraction procedure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service