Skip to Content
Merck
All Photos(1)

Key Documents

51492C

SAFC

Glasgow′s Modified Eagle′s Medium

Synonym(s):

Glasgow Minimum Essential Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

GMEM, with 800 mg/L glycine, with 2750 mg/L sodium bicarbonate, with 110 mg/L sodium pyruvate, without ferric nitrate, without L-glutamine
for research or for further manufacturing use

sterility

sterile

form

liquid

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

components

L-glutamine: no
sodium pyruvate: 110 mg/L
phenol red: 17 mg/L
NaHCO3: 2750 mg/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Glasgow′s Modified Eagle′s Medium has been used to culture baby hamster kidney (BHK-21) cells. It has also been used to maintain undifferentiated mouse embryonic stem cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generation of Kidney from Pluripotent Stem Cells via Blastocyst Complementation
Jo-Ichi Usui
The American Journal of Pathology, 180(6), 2417-2426 (2012)
Inefficient Type I Interferon-Mediated Antiviral Protection of Primary Mouse Neurons Is Associated with the Lack of Apolipoprotein L9 Expression
Marguerite Kreit
Journal of Virology, 88(7), 3874-3884 (2014)
Yira Bermudez et al.
PloS one, 6(5), e20487-e20487 (2011-06-10)
Chronic UV skin exposure leads to epidermal differentiation defects in humans that can be largely restored by pharmacological doses of nicotinic acid. Nicotinic acid has been identified as a ligand for the human G-protein-coupled receptors GPR109A and GPR109B that signal
Rakhi Srivastava et al.
Evidence-based complementary and alternative medicine : eCAM, 2011, 427031-427031 (2011-08-27)
In Indian traditional medicine, Boerhaavia diffusa (punarnava) roots have been widely used for the treatment of dyspepsia, jaundice, enlargement of spleen, abdominal pain and as an anti-stress agent. Pharmacological evaluation of the crude ethanolic extract of B. diffusa roots has
Rebeka R Merson et al.
Aquatic toxicology (Amsterdam, Netherlands), 94(1), 47-55 (2009-06-23)
The aryl hydrocarbon receptor (AHR) mediates the toxic effects of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) and related compounds. In some mammalian cell lines, TCDD induces G1 cell cycle arrest, which depends on an interaction between the AHR and the retinoblastoma tumor suppressor (RB).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service